162965
2-(2-Chloroethoxy)ethanol
99%
Synonym(s):
Diethylene glycol monochlorohydrin
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
124.57
Beilstein:
506015
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
liquid
refractive index
n20/D 1.452 (lit.)
bp
79-81 °C/5 mmHg (lit.)
density
1.18 g/mL at 25 °C (lit.)
functional group
chloro
ether
hydroxyl
SMILES string
OCCOCCCl
InChI
1S/C4H9ClO2/c5-1-3-7-4-2-6/h6H,1-4H2
InChI key
LECMBPWEOVZHKN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
2-(2-Chloroethoxy)ethanol is a chloroalkoxy alcohol commonly used as an organic building block in the synthesis of various pharmaceutical compounds.
Application
2-(2-Chloroethoxy)ethanol was used in the synthesis of o-nitrophenylbromoacetaldehyde bis-2-(2-chloroethoxy)-ethyl acetal, quetiapine (an antipsychotic drug) and 2-(2-azidoethoxy)ethanol.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
194.0 °F - closed cup
Flash Point(C)
90 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
GC method for quantitative determination of residual 2-(2-chloroethoxy) ethanol (CEE) and N-methyl-2-pyrrolidinone (NMP) in quetiapine.
Elzbieta U Stolarczyk et al.
Acta poloniae pharmaceutica, 64(2), 187-189 (2007-08-02)
One-Pot Synthesis of Well-Defined Amphiphilic and Adaptative Block Copolymers via Versatile Combination of ?Click? Chemistry and ATRP.
Mespouille L, et al.
Macromolecular Rapid Communications, 28(22), 2151-2158 (2007)
Abdelreheem Abdelfatah Saddik et al.
Journal of materials chemistry. B, 8(43), 9961-9970 (2020-10-14)
The discovery of crown ethers and their unique interactions with ions make them play a key role in supramolecular chemistry. In this study, we have developed a new type of amphiphilic crown ether (DB18C6, DB24C8)-conjugated phenylalanine dipeptides for the gelation
G C Ellis-Davies et al.
Proceedings of the National Academy of Sciences of the United States of America, 91(1), 187-191 (1994-01-04)
The synthesis and properties of a caged calcium are described. The compound is an ortho-nitrophenyl derivative of EGTA. It is synthesized in 10 steps and with 24% overall yield. The photosensitive chelator, nitrophenyl-EGTA, has a Kd value for Ca2+ of
Yohann Catel et al.
Dental materials : official publication of the Academy of Dental Materials, 37(2), 351-358 (2020-12-29)
To evaluate high refractive index methacrylates as diluents for the formulation of radiopaque esthetic bulk-fill composites. 2-(4-Cumylphenoxy)ethyl methacrylate 1, 2-(2-phenylphenoxy)ethyl methacrylate 2 and 2-[2-(2-phenylphenoxy)ethoxy]ethyl methacrylate 3 were synthesized and characterized by 1H NMR spectroscopy. The reactivity of each monomer was
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service