B3000
1,4-Benzenedimethanol
99%
Synonym(s):
p-Phenylene dicarbinol, p-Phenylenedimethanol, p-Xylene-α,α′-diol, p-Xylylene dialcohol, NSC 5097
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
C6H4(CH2OH)2
CAS Number:
Molecular Weight:
138.16
Beilstein:
2042077
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
powder
bp
138-143 °C/1 mmHg (lit.)
mp
114-118 °C (lit.)
SMILES string
OCc1ccc(CO)cc1
InChI
1S/C8H10O2/c9-5-7-1-2-8(6-10)4-3-7/h1-4,9-10H,5-6H2
InChI key
BWVAOONFBYYRHY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
1,4-Benzenedimethanol acts as a chain extender to prepare dimethylsiloxane-urea-urethane copolymers.
Application
1,4-Benzenedimethanol can be used to prepare:
- Poly(6-methyl-ε-caprolactone), which is a key intermediate for the synthesis of polylactide based thermoplastic elastomers.
- A Highly cross-linked polymer named HCP−BDM (hyper cross-linked polymer-1,4-benzenedimethanol) via Friedel−Crafts alkylation in the presence of Lewis acid.
- Sulfonated polynuclear aromatic resins.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Jinsil Kim et al.
Polymers, 12(4) (2020-04-25)
A functional polyurethane based on the heterocyclic group was synthesized and its self-healing and mechanical properties were examined. To synthesize a heterocyclic polyurethane, a polyol and a heterocyclic compound with di-hydroxyl groups at both ends were blended and the blended
Synthesis of the sulfonated condensed polynuclear aromatic (S-COPNA) resins as strong protonic acids
Tanemura K, et al.
Tetrahedron, 67(6), 1314-1319 (2011)
Polydimethylsiloxane-urea-urethane copolymers with 1, 4-benzenedimethanol as chain extender
Ho T, et al.
Macromolecules, 26, 7029-7036 (1993)
Microporous organic polymers synthesized by self-condensation of aromatic hydroxymethyl monomers
Luo Y, et al.
Polym. Chem., 4(4), 1126-1131 (2013)
Polylactide-poly (6-methyl-ε-caprolactone)-polylactide thermoplastic elastomers
Martello MT and Hillmyer M A
Macromolecules, 44(21), 8537-8545 (2011)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service