A93607
4-Azabenzimidazole
99%
Synonym(s):
1H-Imidazo[4,5-b]pyridine
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
Quality Level
Assay
99%
form
powder
mp
148-151 °C (lit.)
SMILES string
c1cnc2nc[nH]c2c1
InChI
1S/C6H5N3/c1-2-5-6(7-3-1)9-4-8-5/h1-4H,(H,7,8,9)
InChI key
GAMYYCRTACQSBR-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Nucleosides, nucleotides & nucleic acids, 23(8-9), 1313-1320 (2004-12-02)
Nitration of substituted (1-deaza)purines using a mixture of tetrabutylammonium nitrate (TBAN) and trifluoracetic acid anhydride (TFAA) was applied to prepare nitrosubstituted (1-deaza)purines at low temperature. The nitro group influences the system twofold: 1) it activates other substituents towards nucleophilic aromatic
Bioorganic chemistry, 65, 48-56 (2016-02-09)
6-Chloro-2-Aryl-1H-imidazo[4,5-b]pyridine derivatives 1-26 were synthesized and characterized by various spectroscopic techniques. All these derivatives were evaluated for their antiglycation, antioxidant and β-glucuronidase potential followed their docking studies. In antiglycation assay, compound 2 (IC50=240.10±2.50μM) and 4 (IC50=240.30±2.90μM) was found to be
The Journal of organic chemistry, 75(24), 8662-8665 (2010-11-26)
A new method for the synthesis of dihydroimidazo[1,2-a][1,3,5]triazin-4(6H)-ones via copper(I)-catalyzed hydroamination was developed. In addition, for the first time, iodoalkynes were shown to participate in the copper(I)-catalyzed intramolecular hydroamination reaction with exclusive formation of E-isomers.
Journal of medicinal chemistry, 36(9), 1175-1187 (1993-04-30)
A series of 1-benzylbenzimidazole and 3-benzylimidazo[4,5-b]pyridine substituted in the 2-position by an alkanoic or mercaptoalkanoic acid chain was synthesized for evaluation as potential thromboxane A2/prostaglandin H2 (TXA2/PGH2) receptor antagonists. The affinity of each compound for washed human platelet TXA2/PGH2 receptors
The Journal of organic chemistry, 75(24), 8693-8696 (2010-11-13)
Template-free cross-linking of single-stranded DNA bearing octadiynyl side chains at the 7-position of 8-aza-7-deazapurine moieties with bisfunctional azides is reported employing a Cu(I)-catalyzed azide-alkyne "bis-click" reaction. Bis-adducts were formed when the bis-azide:oligonucleotide ratio was 1:1; monofunctionalization occurred when the ratio
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service