Skip to Content
Merck
All Photos(4)

Documents

L2904

Sigma-Aldrich

Lithium tri-tert-butoxyaluminum hydride

97%

Synonym(s):

LTBA, Lithium aluminum-tri-tert-butoxyhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiAlH[OC(CH3)3]3
CAS Number:
Molecular Weight:
254.27
Beilstein:
5796791
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reagent type: reductant

mp

300-319 °C (dec.) (lit.)

SMILES string

[H][Al]([Li])(OC(C)(C)C)(OC(C)(C)C)OC(C)(C)C

InChI

1S/3C4H9O.Al.Li.H/c3*1-4(2,3)5;;;/h3*1-3H3;;;/q3*-1;+2;+1;

InChI key

BYBIDFZFKATBFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Reducing agent
Lithium tri-tert-butoxyaluminum hydride (LTBA) is a stable, mild reducing agent that is used to selectively reduce aldehydes and ketones in the presence of esters. It can also be used to reduce imidoyl chlorides to aldimines and aromatic disulfides to the corresponding thiols.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rapid and selective reduction of functionalized aromatic disulfides with lithium tri-tert-butoxyaluminohydride. A remarkable steric and electronic control. Comparison of various hydride reagents.
Krishnamurthy S and Aimino D
The Journal of Organic Chemistry, 54(18), 4458-4462 (1989)
Remarkably facile reductive opening of tetrahydrofuran and related ethers by lithium tri-tert-butoxyaluminohydride in the presence of triethylborane.
Brown H C, et al.
Journal of the American Chemical Society, 94(5), 1750-1751 (1972)
Facile preparation of polyfluoroalkylated aldimines from polyfluoroalkanoic acids.
Takagi J, et al.
Synthesis, 2007(11), 1624-1628 (2007)
Electrolytic Decarboxylation. 6. A Convenient Synthesis of 3-(cis-3-Hexenyl)-2-cyclopentenone, a Precursor of cis-Jasmone Synthesis.
Torii S, et al.
Bulletin of the Chemical Society of Japan, 55(12), 3947-3948 (1982)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service