Skip to Content
Merck
All Photos(1)

Key Documents

95020

Sigma-Aldrich

Dimethyl vinylphosphonate

≥95% (GC)

Synonym(s):

1-Dimethoxyphosphorylethene, Dimethyl ethenylphosphonate, Vinyl dimethylphosphonate, Vinyl(dimethoxy)oxophosphine, Vinylphosphonic acid dimethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHPO(OCH3)2
CAS Number:
Molecular Weight:
136.09
Beilstein:
1748827
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (GC)

form

liquid

refractive index

n20/D 1.430-1.435

bp

197-202 °C (lit.)

density

1.146 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COP(=O)(OC)C=C

InChI

1S/C4H9O3P/c1-4-8(5,6-2)7-3/h4H,1H2,2-3H3

InChI key

CQCXMYUCNSJSKG-UHFFFAOYSA-N

Application

  • Improving the Corrosion Resistance of Lead in H2SO4 by Addition of Phosphoric and Phosphonic Compounds: This study discusses how Dimethyl Vinylphosphonate enhances lead grid batteries′ corrosion resistance (S Khatbi et al., 2016).
  • Relay Cross Metathesis Reactions of Vinylphosphonates: This publication highlights the reaction of Dimethyl Vinylphosphonate with neat methylimidazole, showcasing a potential route for producing valuable organic salts (RK Malla et al., 2014).
  • Enhanced Flame Retardance of Polyacrylonitrile with Dimethyl Vinylphosphonate: Focuses on improving the flame retardancy of textiles, adding significant value in material safety applications (X Dong et al., 2021).

Other Notes

Vinylphosphonates are versatile reagents undergoing Michael addition giving alkyl phosphonates; synthesis of aziridine-2-phosphonates, useful for preparing α-amino phosphonates

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S. Mirza et al.
Helvetica Chimica Acta, 67, 1562-1562 (1984)
F. Nicotra et al.
Journal of the Chemical Society. Chemical Communications, 5-5 (1984)
N. Minova et al.
Bulletin of the Chemical Society of Japan, 60, 1761-1761 (1987)
J. Zygmunt
Tetrahedron, 41, 4979-4979 (1985)
Amir Shafiee Kisomi et al.
Ultrasonics sonochemistry, 44, 129-136 (2018-04-24)
In this research, a novel Sn(II)-imprinted poly(dimethyl vinylphosphonate) nanopowder (Sn(II)-IPDMVPN) was prepared using Sn2+, dimethyl vinylphosphonate, azobis isobutyronitril and ethylene glycol dimethacrylate as the template, ligand, initiator and cross linker, respectively. The non-imprinted poly(dimethyl vinylphosphonate) nanopowder (NIPDMVPN) was also synthesized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service