Skip to Content
Merck
All Photos(1)

Key Documents

672068

Sigma-Aldrich

Vinylphosphonic acid

≥90% (T)

Synonym(s):

Ethenephosphonic acid, Ethylenephosphonic acid, P-Ethenylphosphonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHP(O)(OH)2
CAS Number:
Molecular Weight:
108.03
Beilstein:
1741622
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥90% (T)

impurities

≤7.0% water

mp

36 °C (Lit. dry VPA) (lit.)

density

1.37 g/mL at 20 °C (lit.)

SMILES string

OP(O)(=O)C=C

InChI

1S/C2H5O3P/c1-2-6(3,4)5/h2H,1H2,(H2,3,4,5)

InChI key

ZTWTYVWXUKTLCP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Vinylphosphonic acid (VPA) can be used as a monomer unit for the synthesis of poly(vinylphosphonic acid) via free radical polymerization. It is also used to develop copolymers of VPA with acrylonitrile, N-isopropylacrylamide, styrene, vinylpyrrolidone, and acrylic and methacrylic acid. These copolymers find potential application in hydrogels, drug delivery, biomimetic mineralization, and polymer electrolyte membranes in fuel cells.
It can also be used as an organic building block to prepare (E)-styryl phosphonic acid derivatives by reacting with various aryl halides via Pd-catalyzed Heck coupling reaction.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

467.6 °F

Flash Point(C)

242 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young Kyung Kim et al.
Biomaterials, 31(25), 6618-6627 (2010-07-14)
The complex morphologies of mineralised collagen fibrils are regulated through interactions between the collagen matrix and non-collagenous extracellular proteins. In the present study, polyvinylphosphonic acid, a biomimetic analogue of matrix phosphoproteins, was synthesised and confirmed with FTIR and NMR. Biomimetic
Synthesis, microstructure, and acidity of poly (vinylphosphonic acid)
Bingol B, et al.
Macromolecular Rapid Communications, 27(20), 1719-1724 (2006)
James D Kretlow et al.
Biomacromolecules, 11(3), 797-805 (2010-02-04)
Stimulus responsive materials hold great promise in biological applications as they can react to changes in physiological stimuli to produce a desired effect. Stimulus responsive macromers designed to respond to temperature changes at or around 37 degrees C and the
Richard D Bertram et al.
Biochemistry, 41(24), 7725-7731 (2002-06-12)
During the past 5 years a great deal of structural and biochemical information has given us a detailed insight into the molecular mechanism of action of the PcrA DNA helicase and challenged previous notions about the molecular mechanism of action
Noh-Seok Kwak et al.
Journal of hazardous materials, 203-204, 213-220 (2012-01-03)
Poly(vinylphosphonic acid-co-methacrylic acid) microbeads were synthesized by suspension polymerization, and their indium adsorption properties were investigated. The obtained microbeads were characterized by Fourier transform infrared (FT-IR) spectroscopy and scanning electron microscopy (SEM). The microbeads were wrinkled spheres, irrespective of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service