Skip to Content
Merck
All Photos(1)

Key Documents

913561

Sigma-Aldrich

Titania nanoparticles

dispersion, 5 wt. % in ethanol, 300 nm particle size

Synonym(s):

TiO2 nanoparticles, TiOxide, Titanium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiO2
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

form

dispersion
nanoparticles

Quality Level

concentration

5 wt. % in ethanol

particle size

300 nm

SMILES string

[Ti](=O)=O

InChI

1S/2O.Ti

InChI key

GWEVSGVZZGPLCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Titania nanoparticles are useful for a wide variety of applications, due to their high refractive index and large bandgap, providing optical properties in the visible spectrum. Applications of titania nanoparticles range from energy to medical applications.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Introduction: titanium dioxide (TiO2) nanomaterials.
Xiaobo Chen et al.
Chemical reviews, 114(19), 9281-9282 (2014-10-09)
Modified titanium oxide (TiO2) nanocomposites and its array of applications: a review.
Ghosh S, et al.
Toxicological & Environmental Chemistry, 97:5, 491-514 (2015)
Mini review on photocatalysis of titanium dioxide nanoparticles and their solar applications.
Lan Y, et al.
Nano Energy, 2(5), 1031-1045 (2013)
Janusz Bogdan et al.
Nanoscale research letters, 12(1), 225-225 (2017-03-30)
Cancer has become a global problem. On all continents, a great number of people are diagnosed with this disease. In spite of the progress in medical care, cancer still ends fatal for a great number of the ill, either as

Articles

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Monodisperse TiO2 nanoparticles fill a gap in commercial sources, offering reliability for various applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service