Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titania nanoparticles
Linear Formula:
TiO
2
CAS Number:
13463-67-7
Products
Brand
Sigma-Aldrich (20)
Physical Form
Quality Segment
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
913928
Titania nanoparticles,
dispersion, 10 wt. % in water, 150 nm particle size
Expand
View Pricing
914576
Titania nanoparticles,
150 nm particle size, dispersion, 5 wt. % in water
Expand
View Pricing
913553
Titania nanoparticles,
300 nm particle size, dispersion, 5 wt. % in water
Expand
View Pricing
914703
Titania nanoparticles,
dispersion, 10 wt. % in water, 900 nm particle size
Expand
View Pricing
914606
Titania nanoparticles,
900 nm particle size, dispersion, 5 wt. % in water
Expand
View Pricing
914835
Titania nanoparticles,
dispersion, 5 wt. % in ethanol, 150 nm particle size
Expand
View Pricing
914177
Titania nanoparticles,
dispersion, 10 wt. % in water, 300 nm particle size
Expand
View Pricing
914673
Titania nanoparticles,
dispersion, 10 wt. % in ethanol, 150 nm particle size
Expand
View Pricing
914967
Titania nanoparticles,
dispersion, 10 wt. % in ethanol, 900 nm particle size
Expand
View Pricing
914320
Titania nanoparticles,
dispersion, 5 wt. % in water, 500 nm particle size
Expand
View Pricing
914193
Titania nanoparticles,
dispersion, 10 wt. % in water, 500 nm particle size
Expand
View Pricing
914932
Titania nanoparticles,
300 nm particle size, dispersion, 10 wt. % in ethanol
Expand
View Pricing
914452
Titania nanoparticles,
dispersion, 10 wt. % in water, 700 nm particle size
Expand
View Pricing
914061
Titania nanoparticles,
dispersion, 5 wt. % in water, 700 nm particle size
Expand
View Pricing
913677
Titania nanoparticles,
dispersion, 10 wt. % in ethanol, 500 nm particle size
Expand
View Pricing
914355
Titania nanoparticles,
dispersion, 5 wt. % in ethanol, 900 nm particle size
Expand
View Pricing
914118
Titania nanoparticles,
dispersion, 5 wt. % in ethanol, 700 nm particle size
Expand
View Pricing
913561
Titania nanoparticles,
dispersion, 5 wt. % in ethanol, 300 nm particle size
Expand
View Pricing
913820
Titania nanoparticles,
dispersion, 5 wt. % in ethanol, 500 nm particle size
Expand
View Pricing
913952
Titania nanoparticles,
dispersion, 10 wt. % in ethanol, 700 nm particle size
Expand
View Pricing