Skip to Content
Merck
All Photos(1)

Key Documents

87642

Sigma-Aldrich

Tetrakis(trimethylsilyl)silane

≥97.0%

Synonym(s):

2,2-Bis(trimethylsilyl)-1,1,1,3,3,3-hexamethyl-trisilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3Si]4Si
CAS Number:
Molecular Weight:
320.84
Beilstein:
1860786
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0%

form

powder

mp

≥300 °C (lit.)
≥300 °C

SMILES string

C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C

InChI

1S/C12H36Si5/c1-13(2,3)17(14(4,5)6,15(7,8)9)16(10,11)12/h1-12H3

InChI key

BOJSDHZZKKYWAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium; Preparation of lithium bis[(TMS)3silyl]cuprate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.H. Ming et al.
Journal of Organometallic Chemistry, 316, 255-255 (1986)
A.G. Brook et al.
Organometallic Syntheses, vol. 4, 385-385 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service