Skip to Content
Merck
All Photos(2)

Key Documents

454516

Sigma-Aldrich

Tetrakis(trimethylsilyloxy)silane

97%

Synonym(s):

1,1,1,5,5,5-Hexamethyl-3,3-bis[(trimethylsilyl)oxy]trisiloxane, Tetrakis(trimethylsilyl) silicate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3SiO]4Si
CAS Number:
Molecular Weight:
384.84
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.389 (lit.)

bp

103-106 °C/2 mmHg (lit.)

density

0.87 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

InChI

1S/C12H36O4Si5/c1-17(2,3)13-21(14-18(4,5)6,15-19(7,8)9)16-20(10,11)12/h1-12H3

InChI key

VNRWTCZXQWOWIG-UHFFFAOYSA-N

Application

Tetrakis(trimethylsilyloxy)silane (TTMS) is an organosilicon compound used as a precursor to prepare nanostructured organosilicon polymer films by plasma-enhanced chemical vapor deposition (PECVD) at atmospheric pressure. TTMS along with cyclohexane can also be used to synthesize low dielectric constant SiCOH films by PECVD method.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liqin Zhi et al.
Journal of environmental sciences (China), 76, 199-207 (2018-12-12)
Methylsiloxanes are a class of silicone compounds that have been widely used in various industrial processes and personal care products for several decades. This study investigated the spatial distribution of three cyclic methylsiloxanes (D4-D6) and twelve linear methylsiloxanes (L5-L16) in
Zichun Huang et al.
Water research, 185, 116224-116224 (2020-08-08)
Based on wastewater and raw/digested sludge samples from 29 wastewater treatment plants in 25 Chinese cities, the nationwide profiles of cis- and trans- isomers of phenylmethylsiloxanes (P3 and P4) and trifluoropropylmethylsiloxanes (D3F and D4F) were investigated. Calculated with paired influents/sludges
Liqin Zhi et al.
Environmental science & technology (2018-10-20)
Fluorinated methylsiloxanes are modified methylsiloxanes and include tris(trifluoropropyl)trimethylcyclotrisiloxane (D3F) and tetrakis(trifluoropropyl)tetramethylcyclotetrasiloxane (D4F). Here, we report fluorinated methylsiloxanes (D3F and D4F) in surface water and sediment samples collected near a fluorinated methylsiloxane manufacturing plant in Weihai, China. The concentrations of D3F
Characteristics of Plasma Polymerized Low-dielectric Constant SiCOH Films Deposited with Tetrakis (trimethylsilyloxy) silane and Cyclohexane Precursors
Kim H, et al.
Bulletin of the Korean Chemical Society,, 35(10), 2941-2944 (2014)
Tetrakis (trimethylsilyloxy) silane for nanostructured SiO2-like films deposited by PECVD at atmospheric pressure
Schafer, J, et al.
Surface and Coatings Technology, 295, 112-118 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service