Skip to Content
Merck
All Photos(2)

Key Documents

862207

Sigma-Aldrich

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one

purified by recrystallization

Synonym(s):

2-Phenyl-1-4-heteromethylene-5-oxazolone, 2-Phenyl-4-(ethoxymethylene)-2-oxazolinone, 2-Phenyl-4-(ethoxymethylene)oxazolone, 2-Phenyl-4-ethoxymethylene-5-oxazolone, 4-(Ethoxymethylene)-2-phenyl-5(4H)-oxazolone, 4-Ethoxymethylene-2-phenyl oxazolone, 4-Ethoxymethylene-2-phenyl-5-oxazolone, Oxazolone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11NO3
CAS Number:
Molecular Weight:
217.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

purified by

recrystallization

mp

94-96 °C (dec.) (lit.)

functional group

ester
ether
phenyl

storage temp.

2-8°C

SMILES string

CCO\C=C1/N=C(OC1=O)c2ccccc2

InChI

1S/C12H11NO3/c1-2-15-8-10-12(14)16-11(13-10)9-6-4-3-5-7-9/h3-8H,2H2,1H3/b10-8-

InChI key

SJHPCNCNNSSLPL-NTMALXAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one is a heterocyclic compound that belongs to the oxazolone family and serves as an important intermediate in the synthesis of amino acids, peptides, and heterocyclic precursors.

Application

4-Ethoxymethylene-2-phenyl-2-oxazolin-5-one can be used as a reactant to synthesize:
  • Pyrimidinone derivatives with N-carboxymethylbenzamidine.
  • Multi-functionalized 1-azabicycles with acyclic enaminones via formal aza-[3+3] cycloaddition.
It can also be used as a source of alpha-amino acid residue to synthesize N-1,2,3-triazolyl substituted amino acid derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yongyan Shi et al.
Cell death & disease, 11(6), 461-461 (2020-06-17)
Crohn's disease (CD) and ulcerative colitis (UC) actually had different pathological mechanisms, as the former was mainly induced by Th1 and Th17 response and the latter by Th2 response. Our previous study found that oxazolone-induced Th2-mediated colitis could not be
Zenghui Lu et al.
The Journal of organic chemistry, 77(21), 9871-9877 (2012-10-11)
A novel palladium-catalyzed approach for the assembly of 3,4,5-trisubstituted oxazolones has been achieved by the coupling of N-alkynyl tert-butyloxycarbamates with aryl halides and related electrophiles, which involves an oxidative addition followed by oxypalladation/reductive elimination. The reaction provides a convenient access
Joanne C Masterson et al.
Gut, 63(1), 43-53 (2012-11-20)
Eosinophilic oesophagitis (EoE) is a chronic inflammatory condition of the oesophagus with limited treatment options. No previous transgenic model has specifically targeted the oesophageal mucosa to induce oesophageal eosinophilia. We developed a mouse model that closely resembles EoE by utilising
Nicolas Pradeille et al.
Chemistry & biodiversity, 9(11), 2528-2558 (2012-11-20)
The total syntheses of hypomurocin A3 and hypomuricin A5 (HM A3 and HM A5, resp.) in solution phase are described. These syntheses have been successfully achieved by applying the 'azirine/oxazolone method' to introduce the two Aib-Pro units into the backbone
Torsten Olszak et al.
Nature, 509(7501), 497-502 (2014-04-11)
The mechanisms by which mucosal homeostasis is maintained are of central importance to inflammatory bowel disease. Critical to these processes is the intestinal epithelial cell (IEC), which regulates immune responses at the interface between the commensal microbiota and the host.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service