Skip to Content
Merck
All Photos(1)

Key Documents

855294

Sigma-Aldrich

8-Aminooctanoic acid

99%

Synonym(s):

ω-Aminocaprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)7COOH
CAS Number:
Molecular Weight:
159.23
Beilstein:
1756493
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

mp

194 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

NCCCCCCCC(O)=O

InChI

1S/C8H17NO2/c9-7-5-3-1-2-4-6-8(10)11/h1-7,9H2,(H,10,11)

InChI key

UQXNEWQGGVUVQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun-Mi Kim et al.
Nuclear medicine and biology, 36(4), 371-378 (2009-05-09)
c-Met is a receptor tyrosine kinase involved in tumor cell growth, invasion, metastases and angiogenesis. Overexpression of c-Met is frequently observed in several tumor types. Here, we report the in vitro cell-binding properties and biodistribution and SPECT/CT imaging in glioma
Jeffrey M Craft et al.
Nuclear medicine and biology, 39(5), 609-616 (2012-01-21)
Bombesin (BN) is an amphibian peptide that binds to the gastrin-releasing peptide receptor (GRPR). It has been demonstrated that BN analogues can be radiolabeled for potential diagnosis and treatment of GRPR-expressing malignancies. Previous studies have conjugated various chelators to the
A J Dobson et al.
Acta crystallographica. Section C, Crystal structure communications, 54 ( Pt 7), 969-972 (1998-09-18)
The title acid, 8-aminooctanoic acid, C8H17NO2, crystallized in the centrosymmetric space group P2(1)/n in the zwitterionic form. The three H atoms involved in hydrogen bonding are ordered. The five intermolecular N-H...O hydrogen bonds have N...O distances ranging from 2.752 (2)
T P Hicks et al.
Neuroscience letters, 57(3), 295-299 (1985-06-24)
The 8-carbon chain analogue of gamma-aminobutyric acid (GABA), omega-aminocaprylic acid (omega-AC), was administered to single cortical neurones of rat by microiontophoresis and its effect on cell firing compared to the effect elicited by GABA. The spontaneous and glutamate-elicited firing rates
Jered C Garrison et al.
Bioconjugate chemistry, 19(9), 1803-1812 (2008-08-21)
The high incidence of BB2 receptor (BB2r) expression in various cancers has prompted investigators to pursue the development of BB2r-targeted agents for diagnostic imaging, chemotherapy, and radiotherapy. Development of BB2r-targeted agents, based on the bombesin (BBN) peptide, has largely involved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service