Skip to Content
Merck
All Photos(1)

Key Documents

798142

Sigma-Aldrich

Fluorescent nanodiamond

Nitrogen vacancy ~1.5ppm NV centers, 100 nm avg. part. size, 1 mg/mL in deionized water

Synonym(s):

Diamond nanopowder, Nanodiamonds

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C
UNSPSC Code:
12352302
NACRES:
NA.23

form

suspension

Quality Level

composition

Nitrogen vacancy, ~1.5ppm NV centers

concentration

1 mg/mL in deionized water

avg. part. size

100 nm

Looking for similar products? Visit Product Comparison Guide

General description

Fluorescent nanodiamonds (FNDs) can be prepared by subjecting synthetic diamond nanocrystallites to helium ions. FND is biocompatible and its surface can be easily functionalized. It is a very useful tool in bioimaging mainly because the photons emitted from the defect centers of FND upon laser excitation can easily penetrate the tissue. The emission from FNDs is extremely stable. The average size of the FNDs is 100 nm.
In fluorescent nanodiamonds, the nitrogen vacancy centers in the diamond lattice provide the fluorescence; unlike the quantum dots or organic dyes, the color centers in these don′t photobleach or photoblink. These outstanding properties of fluorescent nanodiamonds along with their biocompatibility, large surface area etc. make them promising as bioimaging probes.
Product can be functionalized with carboxylic acid, hydroxyl, ketones, ethers, and/or hydrogen.

Application

Potential uses of FNDs are as in vivo nanoprobe for bioimaging, cell labelling and cell tracking.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly Fluorescent Nanodiamonds Protein-Functionalized for Cell Labeling and Targeting
B-M Chang, H-S Lin, L-J Su, et al.
Advances in Functional Materials, 23(46), 5737-5745 (2013)
Nanodiamonds for optical bioimaging
Y. Y. Hui, C-L Cheng, and H-C Chang
Journal of Physics D: Applied Physics, 43(37), 374021-374021 (2010)
The Exocytosis of Fluorescent Nanodiamond and Its Use as a Long-Term Cell Tracker
Fang CY, et al.
Small, 7(23), 3363-3370 (2011)
Mass production and dynamic imaging of fluorescent nanodiamonds
Chang YR, et al.
Nature Nanotechnology, 3(5), 284-288 (2008)

Articles

Fluorescent Nanodiamond Particles (FNDs)- Find properties and applications of nanodiamond particles.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

Fluorescent Nanodiamonds and biofunctionalization strategies are detailed for imaging and bioconjugation applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service