Skip to Content
Merck
All Photos(1)

Key Documents

779202

Sigma-Aldrich

Ethylene oxide solution

2.5-3.3 M in THF

Synonym(s):

Dimethylene oxide, Epoxyethane, Ethyleneoxy, Oxacyclopropane, Oxidoethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O
CAS Number:
Molecular Weight:
44.05
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solution

reaction suitability

reaction type: C-C Bond Formation

concentration

2.5-3.3 M in THF

functional group

ether

SMILES string

C1CO1

InChI

1S/C2H4O/c1-2-3-1/h1-2H2

InChI key

IAYPIBMASNFSPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethylene oxide solution can be used for the conversion of ortho-carborane to (2′-hydroxyethyl)-1,2-dicarba-closo-dodecaborane, a key step in the synthesis of ortho-carborane-modified N-alkyl-deoxynojirimycin (DNMs).

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1B - Repr. 1B - Skin Corr. 1 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ortho?Carborane?Modified N?Substituted Deoxynojirimycin.
Hoogendoorn S
European Journal of Organic Chemistry, 2015(20), 4437-4446 (2015)
Matthieu Gatineau et al.
American journal of veterinary research, 73(10), 1665-1669 (2012-09-28)
To compare effects of sterilization with hydrogen peroxide gas plasma (HPGP), ethylene oxide, and steam on bioadhesive properties of nylon and polyethylene lines used for stabilization of canine stifle joints. Samples of a 36.3-kg test nylon leader line, 57.8-kg test
Filippo Doria et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(1), 78-81 (2012-12-06)
Caught in the oxirane: Naphthalene diimides conjugated to a quinone methide and an oxirane have been synthesized and investigated as selective DNA G-quadruplex alkylating agents. The oxirane derivative generates a stable adduct with a G-quadruplex and shows selective alkylation of
Rasmus Bodvik et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13562-13569 (2012-08-31)
Temperature effects on the viscosity and aggregation behavior of aqueous solutions of three different cellulose ethers--methylcellulose (MC), hydroxypropylmethylcellulose (HPMC), and ethyl(hydroxyethyl)cellulose (EHEC)--were investigated using viscosity and dynamic light scattering measurements as well as cryo-TEM. In all cases, increasing temperature reduces
Guangzhi Gu et al.
Biomaterials, 34(1), 196-208 (2012-10-17)
By taking advantage of the dramatically upregulated expression of matrix metalloproteinases MMP-2 and MMP-9 in glioblastomas and the powerful transport ability of low molecular weight protamine (LMWP), we constructed an activatable low molecular weight protamine (ALMWP) and conjugated it to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service