Skip to Content
Merck
All Photos(1)

Key Documents

48838

Supelco

Ethylene oxide solution

certified reference material, 50 mg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4O
CAS Number:
Molecular Weight:
44.05
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

50 mg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

single component solution

storage temp.

2-8°C

SMILES string

C1CO1

InChI

1S/C2H4O/c1-2-3-1/h1-2H2

InChI key

IAYPIBMASNFSPL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find a list of available reference materials for compounds listed in 10/2011 here


Ethylene oxide is a toxic and flammable liquid, which can be obtained via vapor phase oxidation of ethylene. It can be used in the synthesis of ethylene glycol, glycol ethers, surfactants and ethanolamines. It is an industrially important chemical, that can be used as fumigant to sterilize dental and medical equipments.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Flam. Liq. 2 - Muta. 1B - Repr. 1B - Skin Corr. 1 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mannan S
Lees' Loss Prevention in the Process Industries: Hazard Identification, Assessment and Control (2004)
Harte J, et al.
Toxics A to Z: A Guide to Everyday Pollution Hazards (1991)
Matthieu Gatineau et al.
American journal of veterinary research, 73(10), 1665-1669 (2012-09-28)
To compare effects of sterilization with hydrogen peroxide gas plasma (HPGP), ethylene oxide, and steam on bioadhesive properties of nylon and polyethylene lines used for stabilization of canine stifle joints. Samples of a 36.3-kg test nylon leader line, 57.8-kg test
Filippo Doria et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(1), 78-81 (2012-12-06)
Caught in the oxirane: Naphthalene diimides conjugated to a quinone methide and an oxirane have been synthesized and investigated as selective DNA G-quadruplex alkylating agents. The oxirane derivative generates a stable adduct with a G-quadruplex and shows selective alkylation of
L Ehrenberg et al.
Mutation research, 330(1-2), 41-54 (1995-08-01)
Data for relationships between in vivo doses inferred from levels of hemoglobin (Hb) or DNA adducts and administered (by inhalation or injection) doses of ethylene oxide (EO) in mice, rats and humans are reviewed. At low absorbed doses or dose

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service