Skip to Content
Merck
All Photos(1)

Documents

74750

Sigma-Aldrich

Octadecylamine

≥99.0% (GC)

Synonym(s):

ODA, 1-Aminooctadecane, Stearylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17NH2
CAS Number:
Molecular Weight:
269.51
Beilstein:
636111
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

10 mmHg ( 72 °C)

Quality Level

Assay

≥99.0% (GC)

form

powder

bp

232 °C/32 mmHg (lit.)

mp

50-52 °C (lit.)
52-55 °C

SMILES string

CCCCCCCCCCCCCCCCCCN

InChI

1S/C18H39N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h2-19H2,1H3

InChI key

REYJJPSVUYRZGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Octadecylamine (ODA) is used:
  • To induce hydrophobicity in nanodiamond (ND) powders.
  • In the surface modification of graphite and fullerenes.
  • As a dual source of carbon and nitrogen in the synthesis of N-doped carbon nanotubes (CNTs).
  • To synthesize a single-chain cationic surfactant, bis(amidoethylcarbamoylethyl) octadecylamine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Target Organs

Liver,Gastrointestinal tract,Immune system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wet chemistry route to hydrophobic blue fluorescent nanodiamond.
Mochalin VN and Gogotsi Y
Journal of the American Chemical Society, 131(13), 4594-4595 (2009)
Micro-structural, electron-spectroscopic and field-emission studies of carbon nitride nanotubes grown from cage-like and linear carbon sources.
Ghosh K, et al.
Carbon, 47(6), 1565-1575 (2009)
Direct solvent-free amination of closed-cap carbon nanotubes: a link to fullerene chemistry.
Basiuk EV, et al.
Nano Letters, 4(5), 863-866 (2004)
Graphite oxide: chemical reduction to graphite and surface modification with primary aliphatic amines and amino acids.
Bourlinos AB, et al.
Langmuir, 19(15), 6050-6055 (2003)
Influence of pH on the aggregation morphology of a novel surfactant with single hydrocarbon chain and multi-amine headgroups.
Wang W, et al.
The Journal of Physical Chemistry B, 112(5), 1409-1413 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service