Skip to Content
Merck
All Photos(2)

Documents

734071

Sigma-Aldrich

Di-tert-butylphosphine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H19OP
CAS Number:
Molecular Weight:
162.21
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling

mp

74-82 °C

functional group

phosphine oxide

SMILES string

CC(C)(C)[PH](=O)C(C)(C)C

InChI

1S/C8H19OP/c1-7(2,3)10(9)8(4,5)6/h10H,1-6H3

InChI key

RYLNFGJYRKJXQE-UHFFFAOYSA-N

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Sol. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service