Skip to Content
Merck
All Photos(3)

Key Documents

725145

Sigma-Aldrich

Lithium cobalt phosphate

greener alternative

powder, ≥99% (trace metals analysis)

Synonym(s):

Cobalt lithium phosphate, LCP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCoPO4
CAS Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

Quality Level

Assay

≥99% (trace metals analysis)

form

powder

mol wt

Mw 160.85 g/mol

composition

LiCoPO4

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

density

3.65 g/cm3 (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[Co++].[O-]P([O-])([O-])=O

InChI

1S/Co.Li.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q+2;+1;/p-3

InChI key

SBWRUMICILYTAT-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Lithium cobalt phosphate (LCP) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

LCP belongs to the class of orthorhombic antiferromagnetic materials that is majorly used as a high voltage cathode. It can be synthesized by hydrothermal methods and utilized in the fabrication of Li-Ion batteries.

Features and Benefits

Cathode Materials for High Energy Density Li Ion Rechargeable Batteries; sub-micron particle size results in increased surface area of electrodes. The composition listed allows the preparation of battery electrodes with enhanced performance and durability.

Li-Batteries, material for electrodes design and manufacturing

Legal Information

Product of Engi-Mat Co.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amine, K.; Yasuda, H.; Yamachi, M.
Electrochemical and Solid-State Letters, 3, 178-178 (2000)
Changes in electronic structure between cobalt and oxide ions of lithium cobalt phosphate as 4.8-V positive electrode material
Nakayama M, et al.
Chemistry of Materials, 16(18), 3399-3401 (2004)
Molecular and structural characterization of some violet phosphate pigments for their non-invasive identification in modern paintings.
C Anselmi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 173, 439-444 (2016-10-06)
Hydrothermal synthesis of LiCoPO4 cathode materials for rechargeable lithium ion batteries
Huang X, et al.
Materials Letters, 59(5), 578-582 (2005)
Kornev, I.; Rivera, J. -P.; Gentil, S.; Jansen, A. G. M.;
Physica B: Condensed Matter, 271, 304-304 (1999)

Articles

Nanomaterials for Energy Storage in Lithium-ion Battery Applications

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service