Skip to Content
Merck
All Photos(1)

Key Documents

710172

Sigma-Aldrich

Dithieno[3,2-b:2′,3′-d]thiophene

97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4S3
CAS Number:
Molecular Weight:
196.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97% (HPLC)

form

powder

mp

65-70 °C

SMILES string

c1cc2sc3ccsc3c2s1

InChI

1S/C8H4S3/c1-3-9-7-5(1)11-6-2-4-10-8(6)7/h1-4H

InChI key

VGWBXRXNERKBSJ-UHFFFAOYSA-N

General description

Dithieno[3,2-b:2′,3′-d]thiophene (DTT) is a conductive material that acts as a donor building block for synthesizing a variety of optoelectronic materials. It can be used as an active layer with high charge mobility and environmental stability for development of organic electronic devices.

Application

DTT can be used for a wide range of applications which include electro-luminescence, organic thin film transistors, photochromism, and organic photovoltaic (OPV) solar cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

247.1 °F - closed cup

Flash Point(C)

119.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly luminescent p-conjugated dithienometalloles: photophysical properties and their application in organic light-emitting diodes
Kondo, R.;Yasuda, T.; Adachi, C.; et al.
Journal of Materials Chemistry, 22, 16810-16816 (2012)
Reduced Band Gap Dithieno [3, 2-b: 2 `, 3 `-d] pyrroles: New n-Type Organic Materials via Unexpected Reactivity.
Pappenfus TM, et al.
Organic Letters, 10(8), 1553-1556 (2008)
Novel copolymers incorporating dithieno [3, 2-b: 2?, 3?-d] thiophene moieties for air-stable and high performance organic field-effect transistors.
Lu K, et al.
Journal of Materials Chemistry, 18(29), 3426-3432 (2008)
A new reaction of P4S10 and Lawesson's reagent; a new method for the synthesis of dithieno [3, 2-b; 2?, 3?-d] thiophenes.
Ertas E and Ozturk T
Tetrahedron Letters, 45(17), 3405-3407 (2004)
Improved synthesis of dithieno [3, 2-b: 2?, 3?-d] thiophene (DTT) and derivatives for cross coupling.
Frey J, et al.
Chemical Communications (Cambridge, England), 10(20), 2424-2425 (2002)

Articles

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service