Skip to Content
Merck
All Photos(1)

Key Documents

241636

Sigma-Aldrich

2,2′-Bithiophene

99%

Synonym(s):

2,2′-Bithienyl, 2,2′-Dithienyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6S2
CAS Number:
Molecular Weight:
166.26
Beilstein:
3039
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

bp

260 °C (lit.)

mp

32-33 °C (lit.)

SMILES string

c1csc(c1)-c2cccs2

InChI

1S/C8H6S2/c1-3-7(9-5-1)8-4-2-6-10-8/h1-6H

InChI key

OHZAHWOAMVVGEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2′-Bithiophene is an electron transporting material with the π-electrons present in the system that facilitate charge mobility.

Application

2,2′-Bithiophene can be polymerized to form poly(2,2′-Bithiophene) which can be electrodeposited on indium tin oxide (ITO) substrates for the fabrication of electrochromic devices. It can also be used in the formation of electrode material for the development of supercapacitors.
Substrate used in a rhodium-catalyzed C-H arylation of heteroarenes with aryl iodides.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5, 5 `-Bis (dimesitylboryl)-2, 2 `-bithiophene and 5, 5 ``-bis (dimesitylboryl)-2, 2 `: 5 `, 2 ``-terthiophene as a novel family of electron-transporting amorphous molecular materials.
Noda T and Shirota Y
Journal of the American Chemical Society, 120(37), 9714-9715 (1998)
Weiying He et al.
Nature communications, 9(1), 3866-3866 (2018-09-27)
Nickel-catalyzed catalyst transfer polycondensation (CTP) of thiophenes is an efficient strategy for the controlled synthesis of polythiophenes. However, a detailed view of its reaction mechanism has remained elusive with unresolved questions regarding the geometry and bonding of critical Ni(0) thiophene intermediates.
Electrode material dependent p-or n-like thermoelectric behavior of single electrochemically synthesized poly (2, 2?-bithiophene) layer?application to thin film thermoelectric generator.
Kublitski J, et al.
Journal of Solid State Electrochemistry, 20(8), 2191-2196 (2016)
Ji He et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 16(7), 1049-1056 (2017-05-13)
Three D-D-π-A triphenylamine-coumarin sensitizers were designed and synthesized with a triphenylamine and coumarin double donor and a cyanoacrylic acid acceptor. Thiophene, bithiophene and phenylthiophene were used as π-bridges, respectively, and the effects of different π-bridges on the photophysical properties and
Applications of Poly (indole-6-carboxylic acid-co-2, 2?-bithiophene) Films in High-Contrast Electrochromic Devices.
Kuo C, et al.
Coatings, 8(3), 102-102 (2018)

Articles

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

From Form to Function: Molding Porous Materials in Three Dimensions by Colloidal Crystal Templating

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service