Skip to Content
Merck
All Photos(3)

Key Documents

702102

Sigma-Aldrich

Carbon, mesoporous

greener alternative

Synonym(s):

Starbon 800

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

surface area

>500 m2/g , BET

pore size

0.4-0.7 cm3/g mesoporosity
0-0.25 cm3/g microporosity

bp

4827 °C

mp

3654-3697 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Starbon is mesoporous carbon synthesized by the carbonization of expanded mesoporous starch at high temperatures. Amylose and amylopectin polymer chains in starch self assemble to form large mesoporous structures. Starbon materials exhibit tunable surface energies and high volume. It can be readily functionalized. The unique features are hydrophobicity, graphite-like surface and chemical inertness.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Carbon, mesoporous can be used in the fabrication of optoelectronic and energy storage devices such as supercapacitor, and lithium ion batteries. It can also be used in the formation of a hybrid support that acts as a green tool for water purification. It is a surface tunable material that finds usage in chromatography, catalysis and environmental remediation.
Starbons may be used as a stationary phase for liquid chromatography and as solid support for precious metal catalyst. Starbon supported Pd catalyst was used in a model Heck reaction of iodobenzene.

Legal Information

Starbon is a trademark of University of York

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sustainable polysaccharide-derived mesoporous carbons (Starbon as additives in lithium-ion batteries negative electrodes
Kim S, et al.
Journal of Material Chemistry A, 5(46), 24380-24387 (2017)
Monolithic mesoporous graphitic composites as super capacitors: From Starbons to Starenes
Garcia AM, et al.
Journal of Material Chemistry A, 6(3), 1119-1127 (2018)
Mesoporous carbon nitrides: synthesis, functionalization, and applications
Lakhi KS, et al.
Chemical Society Reviews, 46, 72-101 (2017)
A novel biomass-based support (Starbon) for TiO2 hybrid photocatalysts: a versatile green tool for water purification
Colmenares JC, et al.
Royal Society of Chemistry Advances, 3(43), 20186-20192 (2013)
Starbons: Cooking up Nanostructured Mesoporous Materials
Budarin V, et al.
Material Matters, 4(1), 19-19 (2009)

Articles

Our strategy is to synthesize mesoporous carbonaceous materials (“Starbons”) using mesoporous expanded starch as the precursor without the need for a templating agent.

Mesoporous Materials include a range of high surface area porous silicates with applications in gas adsorption, drug delivery, diagnostics and catalysis.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

Mesoporous materials self-assemble from sol-gel precursors and amphiphiles, forming versatile structures for various applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service