Skip to Content
Merck
All Photos(3)

Key Documents

572497

Sigma-Aldrich

Fullerene soot

greener alternative

(as produced)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

bp

500-600 °C (lit.)

density

~1.7 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

General description

Fullerene soot is a combination of fullerenes and carbon blacks. It is produced by vaporizing pure carbon in an inert atmosphere and condensing the vapor, forming a wide range of structures that include fullerenes.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Fullerene soot is a carbonaceous material that can be used in a variety of sustainable applications such as double-layer capacitor (DLC), water purification, hydrogen storage, supercapacitor, and nanoelectronics.
Fullerene soot is a low cost nanomaterial that finds applications in superconductors, photoconductors and semiconductor based industries.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel application of depleted fullerene soot (DFS) as support of catalysts for low-temperature reduction of NO with CO
Spassova I, et al.
Journal of Colloid and Interface Science, 320(1), 186-193 (2008)
Fullerene production in tons and more: from science to industry.
Murayama H, et al.
Fullerenes, Nanotubes, and Carbon Nanostructures, 12(1-2), 1-9 (2005)
Synthesis and search for superconductivity in LiBC.
Bharathi A, et al.
Solid State Communications, 124(10-11), 423-428 (2002)
NEW IMAGING METHODS TO IMPROVE TEXT LEGIBILITY OF OSTRACA
Bearman
Journal of Archaeology of Egypt/Egyptology, 8(2) (2011)
Toluene-insoluble fraction of fullerene-soot as the electrode of a double-layer capacitor
Egashira M, et al.
Journal of Power Sources, 148, 116-120 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service