Skip to Content
Merck
All Photos(2)

Key Documents

667412

Sigma-Aldrich

Pentamethylcyclopentadienyltris (acetonitrile)ruthenium(II) hexafluorophosphate

Synonym(s):

[RuCp*(MeCN)3]PF6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N3PRuF6
CAS Number:
Molecular Weight:
504.42
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

storage temp.

2-8°C

SMILES string

[Ru+].CC#N.CC#N.CC#N.F[P-](F)(F)(F)(F)F.CC1C(C)=C(C)C(C)=C1C

InChI

1S/C10H16.3C2H3N.F6P.Ru/c1-6-7(2)9(4)10(5)8(6)3;3*1-2-3;1-7(2,3,4,5)6;/h6H,1-5H3;3*1H3;;/q;;;;-1;+1

InChI key

IYEMDYLFXFMECZ-UHFFFAOYSA-N

General description

Pentamethylcyclopentadienyltris (acetonitrile)ruthenium(II) hexafluorophosphate is an organoruthenium compound. It is used as a homogeneous catalyst in organic synthesis. It enables the hydrosilylation of alkynes, cyclotrimerization of alkynes, cycloaddition reactions, and carbon-carbon or carbon-heteroatom bond-forming reactions.

Application

A versatile catalyst for hydrosilylation of alkynes providing α-vinylsilanes.
Reagent use as catalyst precursors for carbon-carbon or carbon-heteroatom bond forming reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barry M Trost et al.
Journal of the American Chemical Society, 127(50), 17644-17655 (2005-12-15)
The complex [Cp*Ru(MeCN)3]PF6 is shown to catalyze the hydrosilylation of a wide range of alkynes. Terminal alkynes afford access to alpha-vinylsilane products with good regioselectivity. Deuterium labeling studies indicate a clean trans addition process is at work. The same complex
Markovnikov alkyne hydrosilylation catalyzed by ruthenium complexes.
B M Trost et al.
Journal of the American Chemical Society, 123(50), 12726-12727 (2001-12-14)

Articles

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Trost group's protocol yields α-vinylsilanes from terminal acetylenes using [Cp*Ru(MeCN)3]PF6 catalyst and others for hydrosilylation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service