Skip to Content
Merck
All Photos(2)

Key Documents

667234

Sigma-Aldrich

Chloro(pentamethylcyclopentadienyl)(cyclooctadiene)ruthenium(II)

Synonym(s):

Cp*RuCl(cod), 1,5-Cyclooctadiene, ruthenium complex, Chloro(1,5-cyclooctadiene)(η5-pentamethylcyclopentadienyl)ruthenium, Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H27ClRu
CAS Number:
Molecular Weight:
379.93
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

core: ruthenium
reagent type: catalyst

mp

143-147 °C

storage temp.

−20°C

SMILES string

Cl[Ru].C1CC=CCCC=C1.C[C]2[C](C)[C](C)[C](C)[C]2C

InChI

1S/C10H15.C8H12.ClH.Ru/c1-6-7(2)9(4)10(5)8(6)3;1-2-4-6-8-7-5-3-1;;/h1-5H3;1-2,7-8H,3-6H2;1H;/q;;;+1/p-1/b;2-1-,8-7-;;

InChI key

MQMQNIQJGNBEMG-ONEVTFJLSA-M

Application

Chloro(pentamethylcyclopentadienyl)(cyclooctadiene)ruthenium(II) is a homogeneous catalyst for the formation of carbon-carbon and carbon-heteroatom bonds.
It can be used:
  • To catalyze cyclotrimerization of alkynylboronates, propargyl alcohols, and terminal alkynes to form arylboronate, which in turn undergoes palladium(II)-catalyzed carbonylation to form highly substituted phthalides.
  • To catalyze C-C coupling of norbornenes and norbornadiene with alkynes to form [2 + 2] cycloadducts.
  • In combination with 2-diphenylphosphinoethylamine-potassium tertiary butoxide to form a ternary catalyst system that can catalyze fast racemization of chiral non-racemic sec-alcohols.
  • To synthesize new organoruthenium complexes with phosphorus-based ligands such as bis(phosphino)amines.
  • To catalyze the addition of organic disulfides to alkenes leading to vic-dithioethers.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The versatility of molecular ruthenium catalyst RuCl(COD)(C5Me5)
D'erien S and Dixneuf PH
Journal of Organometallic Chemistry, 689(8), 1382-1392 (2004)
Cp*RuCl-catalyzed formal intermolecular cyclotrimerization of three unsymmetrical alkynes through a boron temporary tether: Regioselective four-component coupling synthesis of phthalides
Yamamoto Y, et al.
Journal of the American Chemical Society, 127(26), 9625-9631 (2005)
First Transition-Metal Complex Catalyzed Addition of Organic Disulfides to Alkenes Enables the Rapid Synthesis of v icinal-Dithioethers
Kondo T, et al.
Journal of the American Chemical Society, 121(2), 482-483 (1999)
[2+ 2] Cycloaddition of norbornenes with alkynes catalyzed by ruthenium complexes
Mitsudo T-A, et al.
Angewandte Chemie (International ed. in English), 33(5), 580-581 (1994)
Rapid racemization of chiral non-racemic sec-alcohols catalyzed by (η5-C5(CH3)5)Ru complexes bearing tertiary phosphine-primary amine chelate ligands
Ito M, et al.
Tetrahedron Letters, 44(40), 7521-7523 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service