Skip to Content
Merck
All Photos(3)

Key Documents

661910

Sigma-Aldrich

(R)-(−)-2-(tert-Butyl)-3-methyl-4-imidazolidinone trifluoroacetic acid

96%

Synonym(s):

(R)-(−)-2-(tert-Butyl)-3-methyl-4-oxoimidazolidinium trifluoroacetate, (R)-2-(tert-Butyl)-3-methyl-4-imidazolidinone trifluoroacetic acid salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N2O3F3
CAS Number:
Molecular Weight:
270.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

solid

mp

99-103 °C

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.CN1[C@@H](NCC1=O)C(C)(C)C

InChI

1S/C8H16N2O.C2HF3O2/c1-8(2,3)7-9-5-6(11)10(7)4;3-2(4,5)1(6)7/h7,9H,5H2,1-4H3;(H,6,7)/t7-;/m1./s1

InChI key

HKHKOEMMPVPVOS-OGFXRTJISA-N

Application

Highly selective organocatalyst for 1,3-dipolar addition and Friedel-Crafts alkylation.

Legal Information

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New strategies in organic catalysis: the first enantioselective organocatalytic Friedel-Crafts alkylation.
N A Paras et al.
Journal of the American Chemical Society, 123(18), 4370-4371 (2001-07-18)
Jen, W. S. et al
Journal of the American Chemical Society, 122, 9874-9874 (2000)

Articles

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service