Skip to Content
Merck
All Photos(1)

Key Documents

66181

Sigma-Aldrich

N-Succinimidyl N-methylcarbamate

≥97.0% (N), for peptide synthesis

Synonym(s):

MIC substitute, Methyl isocyanate substitute

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O4
CAS Number:
Molecular Weight:
172.14
Beilstein:
1531863
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Product Name

N-Succinimidyl N-methylcarbamate, ≥97.0% (N)

Quality Level

Assay

≥97.0% (N)

form

solid

application(s)

peptide synthesis

functional group

amine
imide

storage temp.

2-8°C

SMILES string

CNC(=O)ON1C(=O)CCC1=O

InChI

1S/C6H8N2O4/c1-7-6(11)12-8-4(9)2-3-5(8)10/h2-3H2,1H3,(H,7,11)

InChI key

XMNGSPOWUCNRMO-UHFFFAOYSA-N

Other Notes

Safe and crystalline chemical equivalent of the toxic methyl isocyanate, MIC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K. Takeda et al.
Tetrahedron Letters, 24, 4569-4569 (1983)
J Martinez et al.
Journal of medicinal chemistry, 25(2), 178-182 (1982-02-01)
A practical and convenient method for synthesizing antitumor compounds, N-alkyl-N-nitrosoureas, regioselectively nitrosated on the nitrogen atom bearing the alkyl group is proposed. N-Alkyl-N-nitrosocarbamates are interesting intermediates in these syntheses and yield, by reaction with amino compounds, the regioselectively nitrosated N-alkyl-N-nitrosoureas.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service