Skip to Content
Merck
All Photos(2)

Key Documents

A8060

Sigma-Aldrich

Acrylic acid N-hydroxysuccinimide ester

≥90%, for peptide synthesis

Synonym(s):

N-Acryloxysuccinimide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO4
CAS Number:
Molecular Weight:
169.13
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Product Name

Acrylic acid N-hydroxysuccinimide ester, ≥90%

Assay

≥90%

form

powder

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C=CC(=O)ON1C(=O)CCC1=O

InChI

1S/C7H7NO4/c1-2-7(11)12-8-5(9)3-4-6(8)10/h2H,1,3-4H2

InChI key

YXMISKNUHHOXFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acrylic acid N-hydroxysuccinimide ester (NAS) is a general acrylating agent that is used in the synthesis of:
  • Acrylated hyaluronic acid (HA) hydrogel for bone regeneration applications.
  • Injectable, biodegradable, and thermosensitive copolymers for tissue engineering.
  • Imidazole-based biocompatible, aqueous quantum dots (QDs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein-reactive, thermoresponsive copolymers with high flexibility and biodegradability.
Guan J, et al.
Biomacromolecules, 9(4), 1283-1292 (2008)
Bone regeneration using hyaluronic acid-based hydrogel with bone morphogenic protein-2 and human mesenchymal stem cells.
Kim J, et al.
Biomaterials, 28(10), 1830-1837 (2007)
Farshad Oveissi et al.
Biomaterials science, 7(3), 926-937 (2018-12-07)
Novel hydrogels with tunable mechanical properties similar to human soft tissue have increasing applications in biomedicine, soft robotics, and biocompatible electronics. However, most of these materials require multiple-step fabrication, are not robust, and compromise bioactivity. Thus, aiming to address these
Fred Y Shen et al.
Nature communications, 11(1), 4632-4632 (2020-09-17)
Mapping neuroanatomy is a foundational goal towards understanding brain function. Electron microscopy (EM) has been the gold standard for connectivity analysis because nanoscale resolution is necessary to unambiguously resolve synapses. However, molecular information that specifies cell types is often lost
Compact biocompatible quantum dots via RAFT-mediated synthesis of imidazole-based random copolymer ligand.
Liu W, et al.
Journal of the American Chemical Society, 132(2), 472-483 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service