Skip to Content
Merck
All Photos(1)

Key Documents

654027

Sigma-Aldrich

Triarylsulfonium hexafluoroantimonate salts, mixed

50 wt. % in propylene carbonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

concentration

50 wt. % in propylene carbonate

refractive index

n20/D 1.502

bp

>220 °C

density

1.410 g/mL at 25 °C

SMILES string

F[Sb-](F)(F)(F)(F)F.F[Sb-](F)(F)(F)(F)F.F[Sb-](F)(F)(F)(F)F.S(c1ccccc1)c2ccc(cc2)[S+](c3ccccc3)c4ccccc4.S(c5ccc(cc5)[S+](c6ccccc6)c7ccccc7)c8ccc(cc8)[S+](c9ccccc9)c%10ccccc%10

InChI

1S/C36H28S3.C24H19S2.18FH.3Sb/c1-5-13-31(14-6-1)38(32-15-7-2-8-16-32)35-25-21-29(22-26-35)37-30-23-27-36(28-24-30)39(33-17-9-3-10-18-33)34-19-11-4-12-20-34;1-4-10-20(11-5-1)25-21-16-18-24(19-17-21)26(22-12-6-2-7-13-22)23-14-8-3-9-15-23;;;;;;;;;;;;;;;;;;;;;/h1-28H;1-19H;18*1H;;;/q+2;+1;;;;;;;;;;;;;;;;;;;3*+5/p-18

InChI key

YASCSMWRFXPFOO-UHFFFAOYSA-A

Looking for similar products? Visit Product Comparison Guide

General description

Triarylsulfonium hexafluoroantimonate salts, mixed are photoinitiators that are used in the cationic polymerization. These photoinitiators are most effective in the range of 210−350 nm. They are majorly used to initiate epoxy resins.

Application

  • 3D Printing Fabrication of Amorphous Thermoelectric Materials: This study explores the use of triarylsulfonium hexafluoroantimonate salts as cationic photoinitiators in the fabrication of thermoelectric materials with ultralow thermal conductivity, highlighting their role in the processing of thermoelectric polymers (M He et al., 2015).
  • Onium Salts for Cationic Polymerization: Discusses the high yields and functionalities of mixed triarylsulfonium salts in cationic polymerization processes, providing a pathway to synthesize asymmetrical triarylsulfonium salts (DIR Taschner, 2023).
  • Cationic UV-curing of Epoxidized Biobased Resins: This paper investigates the dual UV and visible light curing of biobased resins using triarylsulfonium hexafluoroantimonate salts, emphasizing its applications in sustainable polymer chemistry (C Noè et al., 2020).
  • Photocurable Epoxy Acrylate Coatings: Examines the dual cationic and radical photocrosslinking of epoxy acrylate coatings using triarylsulfonium hexafluoroantimonate salts, focusing on the properties of hybrid polymer networks (P Bednarczyk et al., 2021).
  • Recent Advances in Monocomponent Visible Light Photoinitiating Systems: This study features a system using triarylsulfonium salt as a photoinitiator for advanced polymerization techniques, demonstrating the versatility of these salts in polymer chemistry (F Dumur, 2023).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>284.0 °F - closed cup

Flash Point(C)

> 140 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A study of epoxy resin-acrylated polyurethane semi-interpenetrating polymer networks
Vabrik R, et al.
Journal of Applied Polymer Science, 68(1), 111-119 (1998)
Photochemistry of triarylsulfonium salts
Dektar JL and Hacker NP
Journal of the American Chemical Society, 112(16), 6004-6015 (1990)
Influence of the initiating mechanism on the cationic photopolymerization of a cycloaliphatic epoxy resin with arylsulfonium salts
Fernandez-Francos X
Journal of Polymer Science Part A: Polymer Chemistry, 45(1), 16-25 (2007)
Kinetics study of the UV-initiated cationic polymerization of cycloaliphatic diepoxide resins.
Voytekunas VY, et al.
European Polymer Journal, 44(11), 3640-3649 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service