548014
Diphenyliodonium hexafluorophosphate
≥98%
Synonym(s):
DPIHFP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C6H5)2I(PF6)
CAS Number:
Molecular Weight:
426.08
Beilstein:
4343861
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Assay
≥98%
mp
140-144 °C (lit.)
SMILES string
F[P-](F)(F)(F)(F)F.[I+](c1ccccc1)c2ccccc2
InChI
1S/C12H10I.F6P/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-7(2,3,4,5)6/h1-10H;/q+1;-1
InChI key
DSSRLRJACJENEU-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Cationic photoinitiator. Photoacid generator.
Packaging
Packaged in amber poly bottles
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Alinne Segatto Pires Altmann et al.
The Angle orthodontist, 87(5), 759-765 (2017-07-08)
The aim of this study was to develop an experimental orthodontic adhesive and evaluate how adding phosphate invert glass containing niobium pentoxide (PIG-Nb) affected the adhesive's properties. PIG-Nb was added at 1, 2.5, and 5 wt% to experimental adhesive (75
C M F Hardy et al.
Dental materials : official publication of the Academy of Dental Materials, 34(9), 1278-1288 (2018-06-03)
To determine the limitations of using light-curable resin-based luting composites (RBLCs) to bond indirect ceramic/resin-composite restorations by measuring light transmittance through indirect restorative materials and the resulting degree of conversion (DC) of the luting-composites placed underneath. Various thicknesses (0-4mm) and
Rizacan Sarikaya et al.
Polymers, 12(9) (2020-09-17)
The inherent degradation property of most dental resins in the mouth leads to the long-term release of degradation by-products at the adhesive/tooth interface. The by-products increase the virulence of cariogenic bacteria, provoking a degradative positive-feedback loop that leads to physicochemical
Monika Topa et al.
Sensors (Basel, Switzerland), 20(11) (2020-05-31)
Novel fluorescent sensors with electron-donating or electron-withdrawing substituents incorporated into a chromophore group based on 2,6-diphenylpyridine were designed and synthesised. The spectroscopic properties of these compounds were studied. Moreover, the positive solvatochromism of 2,6-bis-(4-methylsulphanylphenyl)pyridine (PT-SCH3) in selected solvents was studied
Aurélien Bigot et al.
Journal of the American Chemical Society, 133(35), 13778-13781 (2011-08-19)
A new strategy for the catalytic enantioselective α-arylation of N-acyloxazolidinones with chiral copper(II)-bisoxazoline complexes and diaryliodonium salts is described. The mild catalytic conditions are operationally simple, produce valuable synthetic building blocks in excellent yields and enantioselectivities, and can be applied
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service