Skip to Content
Merck
All Photos(1)

Documents

641065

Sigma-Aldrich

2-Oxopiperazine

97%

Synonym(s):

2-Ketopiperazine, 3,4,5,6-Tetrahydropyrazin-2-ol, 3-Oxopiperazine, Piperazin-2-one, Piperazine-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O
CAS Number:
Molecular Weight:
100.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

136-140 °C (lit.)

SMILES string

O=C1CNCCN1

InChI

1S/C4H8N2O/c7-4-3-5-1-2-6-4/h5H,1-3H2,(H,6,7)

InChI key

IWELDVXSEVIIGI-UHFFFAOYSA-N

Application

2-oxopiperazine is an important building block in medicinal chemistry, since it forms the structural moiety of many natural products and biologically active compounds.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Skeletal diversity by sequential one-pot and stepwise routes using morpholine ester scaffolds"
Ciofi L, et al.
Tetrahedron Letters, 51(48), 6282-6285 (2010)
Kuhan Chandru et al.
Scientific reports, 10(1), 17560-17560 (2020-10-18)
Prebiotic chemists often study how modern biopolymers, e.g., peptides and nucleic acids, could have originated in the primitive environment, though most contemporary biomonomers don't spontaneously oligomerize under mild conditions without activation or catalysis. However, life may not have originated using
"Oligooxopiperazines as nonpeptidic a-helix mimetics"
Tosovska P and Arora.SP
Organic Letters, 12(07), 1588-1591 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service