Skip to Content
Merck
All Photos(3)

Key Documents

637998

Sigma-Aldrich

Vinylboronic anhydride pyridine complex

95%

Synonym(s):

2,4,6-Trivinylcyclotriboroxane pyridine complex, Trivinyl-boroxin pyridine complex, Trivinylcyclotriboroxane pyridine complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9B3O3 · C5H5N
CAS Number:
Molecular Weight:
240.67
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

storage temp.

−20°C

SMILES string

c1ccncc1.C=Cb2ob(C=C)ob(C=C)o2

InChI

1S/C6H9B3O3.C5H5N/c1-4-7-10-8(5-2)12-9(6-3)11-7;1-2-4-6-5-3-1/h4-6H,1-3H2;1-5H

InChI key

YLHJACXHRQQNQR-UHFFFAOYSA-N

Application

Reagent used for
  • Suzuki-Miyaura cross-coupling
  • Stereoselective synthesis via Palladium-catalyzed carboamination
  • Alkyl-connected 2-amino-6-vinylpurine (AVP) crosslinking agent to cytosine base in RNA
  • Kaiser oxime resin-derived palladacycle as a recoverable polymeric precatalyst in Suzuki-Miyaura cross-coupling reactions in aqueous media
  • Kinetic resolution of phosphoryl and sulfonyl esters of binaphthol derivatives via Pd-catalyzed alcoholysis of their vinyl ethers
  • Stereoselective isomerization of N-allyl aziridines into Z-enamines by using rhodium hydride catalysis
  • Kinetic resolution of axially chiral biaryl derivatives via palladium/chiral diamine ligand-catalyzed alcoholysis
  • Transition metal-catalyzed alkenylation of aziridines, cycloaddition and thermal rearrangement reactions
  • Intramolecular Heck reaction strategy for synthesis of functionalized tetrahydroanthracenes

Reagent used for Preparation of
  • BACE-1 inhibitors and SAR of cyclic sulfone hydroxyethylamines
  • Distorted spiropentanes
  • Small molecule bradykinin B2 receptor antagonists in angioedema therapy
  • Enol Ethers
  • Styryl cyclobutanone

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroshi Aoyama et al.
Journal of the American Chemical Society, 127(30), 10474-10475 (2005-07-28)
Palladium-diamine complexes catalyzed kinetic resolution of axially chiral 2,2'-dihydroxy-1,1'-biaryls by alcoholysis of vinyl ethers. The reaction proceeded with high selectivity for various kinds of biaryls. This process is applicable to not only binaphthols but also biphenols, which have been considered
Heinrich Rueeger et al.
Bioorganic & medicinal chemistry letters, 21(7), 1942-1947 (2011-03-11)
This Letter describes the de novo design of non-peptidic hydroxyethylamine (HEA) inhibitors of BACE-1 by elimination of P-gp contributing amide attachments. The predicted binding mode of the novel cyclic sulfone HEA core template was confirmed in a X-ray co-crystal structure.
Shadi Dalili et al.
Organic letters, 7(6), 1161-1164 (2005-03-12)
[reaction: see text] Straightforward methods for palladium-catalyzed alkenylation of aziridines with alkenyl halides and copper-catalyzed alkenylation of aziridines with alkenyl boronic acids have been developed. This methodology offers attractive alternatives to the known methods requiring activated alkenyl halides and acetylenes.
Yosuke Taniguchi et al.
Bioorganic & medicinal chemistry, 18(8), 2894-2901 (2010-03-30)
We have previously reported that the 2-amino-6-vinylpurine (AVP) nucleoside exhibits a highly efficient and selective crosslinking reaction toward cytosine and displayed an improved antisense inhibition in cultured cells. In this study, we further investigated the alkyl-connected AVP nucleoside analogs for
Kinetic resolution of phosphoryl and sulfonyl esters of 1,1'-bi-2-naphthol via Pd-catalyzed alcoholysis of their vinyl ethers
Sakuma, T.; et al.
Tetrahedron, 19, 1593-1599 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service