Skip to Content
Merck
All Photos(1)

Key Documents

608742

Sigma-Aldrich

Sodium nitrate-14N

99.95 atom % 14N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na14NO3
Molecular Weight:
84.99
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

description

15N-depleted

Quality Level

isotopic purity

99.95 atom % 14N

form

solid

mp

306 °C (lit.)

mass shift

depleted

SMILES string

[Na+].[O-][14N+]([O-])=O

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1/i1+0;

InChI key

VWDWKYIASSYTQR-XUGDXVOUSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geraldine Mulley et al.
PloS one, 9(4), e94409-e94409 (2014-04-15)
There has been a recent surge in the use of silver as an antimicrobial agent in a wide range of domestic and clinical products, intended to prevent or treat bacterial infections and reduce bacterial colonization of surfaces. It has been
Anna-Maria Georgoudaki et al.
Immunology and cell biology, 93(6), 581-590 (2015-02-04)
Signaling lymphocytic activation molecule (SLAM) receptors have an important role in the development of immune responses because of their roles, for exampe, in NK cell cytotoxicity and cytokine production by NK, T cells and myeloid cells. The SLAM receptor CD244
Simone Porcelli et al.
Medicine and science in sports and exercise, 47(8), 1643-1651 (2014-11-21)
Dietary nitrate supplementation has been shown to reduce O2 cost of submaximal exercise, improve exercise tolerance, and enhance performance in moderately trained individuals. In contrast, data have been provided that elite athletes do not benefit from nitrate supplementation. The aim
Dong Woog Lee et al.
Nature communications, 6, 7238-7238 (2015-05-27)
Membrane fusion is the core process in membrane trafficking and is essential for cellular transport of proteins and other biomacromolecules. During protein-mediated membrane fusion, membrane proteins are often excluded from the membrane-membrane contact, indicating that local structural transformations in lipid
Daysi Perez-Rea et al.
Analytical and bioanalytical chemistry, 407(15), 4315-4326 (2015-05-01)
We have investigated methods of starch dissolution with the aim of finding an optimum method to completely dissolve starch granules to form a molecularly dissolved starch solution without degradation of the polymers. Glycogen was used as a model molecule for

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service