364592
Carbon-13C dioxide
99.0 atom % 13C, <3 atom % 18O
Synonym(s):
13C Labeled carbon dioxide, 13CO2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
13CO2
CAS Number:
Molecular Weight:
45.00
Beilstein:
3087932
MDL number:
UNSPSC Code:
12352119
PubChem Substance ID:
NACRES:
NA.11
Recommended Products
vapor density
1.52 (vs air)
Quality Level
isotopic purity
99.0 atom % 13C
<3 atom % 18O
Assay
99% (CP)
mp
−78 °C (subl.) (lit.)
mass shift
M+1
storage temp.
room temp
SMILES string
O=[13C]=O
InChI
1S/CO2/c2-1-3/i1+1
InChI key
CURLTUGMZLYLDI-OUBTZVSYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Important basic building block for 13C labeled compounds.
Packaging
All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard products are packaged in a 450 mL carbon steel lecture bottle with brass CGA 110/180 valve. Nominal gas pressures at 21ºC are 20 psig for 1 L and 290 psig for 10 L. These pressures are slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
Recommended products
Stainless steel control valve Z146951 or Z187224 is recommended.
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986
PTFE sealing washers GCA inlet size 180 Z146994
PTFE sealing washers CGA inlet size 110 Z146986
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service