Skip to Content
Merck
All Photos(3)

Key Documents

577782

Sigma-Aldrich

Sodium carbonate decahydrate

99.999% trace metals basis

Synonym(s):

Sal soda, Soda

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2CO3 · 10H2O
CAS Number:
Molecular Weight:
286.14
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

crystalline

impurities

≤15.0 ppm Trace Metal Analysis

SMILES string

O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O

InChI

1S/CH2O3.2Na.10H2O/c2-1(3)4;;;;;;;;;;;;/h(H2,2,3,4);;;10*1H2/q;2*+1;;;;;;;;;;/p-2

InChI key

XYQRXRFVKUPBQN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Sodium carbonate decahydrate (Na2CO3.10H2O) can be used in the preparation of eutectic hydrate salts, which can be used to remove phase change materials for thermal energy storage. It can also be dehydrated to generate cold heat using a chemical heat pump.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heat pump characteristics of sodium carbonate dehydration/hydration system
Matsumura Y and Yoshida K
International Journal of Energy Research, 19(3), 253-261 (1995)
Graphene oxide modified hydrate salt hydrogels: form-stable phase change materials for smart thermal management
Liu Y, et al.
Journal of Material Chemistry A, 4(46), 18134-18143 (2016)
Preparation and thermal properties of Na2CO3 10H2O-Na2HPO4 12H2O eutectic hydrate salt as a novel phase change material for energy storage
Liu Y and Yang Y
Applied Thermal Engineering, 112, 606-609 (2017)
Azadeh Nilghaz et al.
Lab on a chip, 12(1), 209-218 (2011-11-18)
This paper describes the fabrication of microfluidic cloth-based analytical devices (μCADs) using a simple wax patterning method on cotton cloth for performing colorimetric bioassays. Commercial cotton cloth fabric is proposed as a new inexpensive, lightweight, and flexible platform for fabricating
Feng Gu et al.
Bioresource technology, 124, 299-305 (2012-09-20)
Green liquor consists of sodium carbonate and sodium sulfide and is readily available in any kraft mills. The green liquor pretreatment process for bioethanol production was developed for wood chips. This process uses only proven technology and equipment currently used

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service