565881
2,8,9-Triisobutyl-2,5,8,9-tetraaza-1-phosphabicyclo[3.3.3]undecane
97%
Synonym(s):
Triisobutylphosphatrane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H39N4P
CAS Number:
Molecular Weight:
342.50
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.5020 (lit.)
density
0.964 g/mL at 25 °C (lit.)
SMILES string
CC(C)CN1CCN2CCN(CC(C)C)P1N(CC2)CC(C)C
InChI
1S/C18H39N4P/c1-16(2)13-20-10-7-19-8-11-21(14-17(3)4)23(20)22(12-9-19)15-18(5)6/h16-18H,7-15H2,1-6H3
InChI key
WFHPXSHLCFHEIA-UHFFFAOYSA-N
Related Categories
Application
Ligand used in the α-arylation of nitriles with aryl bromides and chlorides, and in the Stille cross-coupling of aryl chlorides.
Together with Pd2(dba)3 (Catalog Number 328774) forms a highly efficient catalyst for one-pot syntheses of trans-4-N,N-diarylaminostilbenes and N,N-diarylaminostyrenes. Catalyst for methanolic cleavage of heteroaromatic carbamates.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
154.9 °F - closed cup
Flash Point(C)
68.3 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tetrahedron Letters, 47, 5645-5645 (2006)
Weiping Su et al.
Organic letters, 6(9), 1421-1424 (2004-04-23)
[reaction: see text] The Pd(2)(dba)(3)/P(i-BuNCH(2)CH(2))(3)N (1d) catalyst system is highly effective for the Stille cross-coupling of aryl chlorides with organotin compounds. This method represents only the second general method for the coupling of aryl chlorides. Other proazaphosphatranes possessing benzyl substituents
A general method for the direct alpha-arylation of nitriles with aryl chlorides.
Jingsong You et al.
Angewandte Chemie (International ed. in English), 42(41), 5051-5053 (2003-11-05)
P(i-BuNCH2CH2)3N: an efficient ligand for the direct alpha-arylation of nitriles with aryl bromides.
Jingsong You et al.
The Journal of organic chemistry, 68(21), 8003-8007 (2003-10-11)
A new catalyst system for the synthesis of alpha-aryl-substituted nitriles is reported. The bicyclic triaminophosphine P(i-BuNCH(2)CH(2))(3)N (1b) serves as an efficient and versatile ligand for the palladium-catalyzed direct alpha-arylation of nitriles with aryl bromides. Using ligand 1b, ethyl cyanoacetate and
Tetrahedron, 61, 9775-9775 (2005)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service