Skip to Content
Merck
Butyl 4-hydroxybenzoate SAJ first grade, ≥99.0%

Butyl 4-hydroxybenzoate

Butyl 4-hydroxybenzoate SAJ first grade, ≥99.0%
Synonym(s):
p-Hydroxybenzoic acid n-butyl ester, Butyl paraben
Linear Formula:
HOC6H4CO2(CH2)3CH3
CAS Number:
Molecular Weight:
194.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing