Skip to Content
Merck
All Photos(1)

Documents

532967

Sigma-Aldrich

Pyridine-d5

≥99.5 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Pentadeuteropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D5N
CAS Number:
Molecular Weight:
84.13
Beilstein:
114377
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

≥99.5 atom % D

Quality Level

Assay

≥99% (CP)

form

solution

contains

0.03 % (v/v) TMS

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

refractive index

n20/D 1.506 (lit.)

pH

8.5 (0.2 g/L)

bp

114.4 °C (lit.)

mp

-41 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1nc([2H])c([2H])c([2H])c1[2H]

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i1D,2D,3D,4D,5D

InChI key

JUJWROOIHBZHMG-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine-d5 is a deuterated NMR solvent useful in NMR-based research and analyses. It has been synthesized by palladium catalyzed H/D (hydrogen/deuterium) exchange reaction between pyridine vapor and heavy water. Infrared and Raman spectra of pyridine-d5 have been recorded in the range 300–4000cm-1.

Application

Pyridine-d5 may be used as a solvent in the 1H NMR based structural analysis of lignin acetates obtained from spruce and birch.

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

62.6 °F

Flash Point(C)

17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Indra Prakash et al.
Natural product communications, 9(8), 1135-1138 (2014-09-23)
We report the isolation and complete structure of an isomer of rebaudioside D, known as rebaudioside D2. This novel steviol glycoside was isolated from a bioconversion reaction of rebaudioside A to rebaudioside D. Rebaudioside D2 possesses a relatively rare 1
Xu Pang et al.
Steroids, 93, 68-76 (2014-12-03)
For the first time, a systematic phytochemical study was performed on the roots of Marsdenia tenacissima. Finally, sixteen new polyoxypregnane glycosides, marstenacissides A1-A7 (1-7) and marstenacissides B1-B9 (8-16), were isolated from M. tenacissima roots. The structures of these new compounds
Xu Pang et al.
Carbohydrate research, 402, 236-240 (2014-12-17)
Five new steroidal saponins (1-5) were isolated from the fermentation broth of total furostanol glycosides from tubers of Dioscorea zingiberensis C.H. Wright incubated with a fungal, Absidia coerulea AS 3.3389, along with known saponins, zingiberensis new saponin (6), deltonin (7)
Yu Shan et al.
Molecules (Basel, Switzerland), 20(11), 20334-20340 (2015-11-17)
A new nortriterpene saponin, 3-O-β-d-glucuronopyranosyl-30-norolean-12,20(29)-dien-23- oxo-28-oic acid, namely bigelovii D (11), was isolated from the hydroalcoholic extract of herbs of Salicornia bigelovii along with 10 known saponins (1-10). Their chemical structures were identified on the basis of spectroscopic analyses including
Shao-Dan Chen et al.
Fitoterapia, 102, 67-73 (2015-02-24)
Five new diarylheptanoids (1-5), along with nine known ones (6-14), were isolated from the rhizomes of Curcuma kwangsiensis. Their structures were established on the basis of spectroscopic analyses. Compounds 1-3 were cyclic diarylheptanoids rarely discovered from C. kwangsiensis. Of all

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service