Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyridine-d
5
Synonym(s):
Pentadeuteropyridine
Empirical Formula (Hill Notation):
C
5
D
5
N
CAS Number:
7291-22-7
Molecular Weight:
84.13
EC Number:
230-720-2
Products
Product Category
solvents (3)
stable isotopes (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
532975
Pyridine-d
5
,
≥99.5 atom % D
Expand
View Pricing
532967
Pyridine-d
5
,
≥99.5 atom % D, contains 0.03 % (v/v) TMS
Expand
View Pricing
177970
Pyridine-d
5
,
"100%", ≥99.96 atom % D
Expand
View Pricing