Skip to Content
Merck
All Photos(1)

Key Documents

532363

Sigma-Aldrich

(S)-(−)-1,2-Epoxybutane

98%

Synonym(s):

(2S)-Ethyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D −10°, neat

refractive index

n20/D 1.386 (lit.)

bp

63 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CC[C@H]1CO1

InChI

1S/C4H8O/c1-2-4-3-5-4/h4H,2-3H2,1H3/t4-/m0/s1

InChI key

RBACIKXCRWGCBB-BYPYZUCNSA-N

Application

(S)-(−)-1,2-Epoxybutane can be used:
  • As a starting material to prepare (+)- and (−)-homononactic acids, which are used as intermediates in the total synthesis of a cyclic antibiotic tetranactin.
  • To prepare a chiral phosphorus synthon, which is applicable in the synthesis of phytoprostane B1 type I.
  • To prepare Eu3+-based precatalysts applicable in the Mukaiyama Aldol reaction in water.

Legal Information

Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.0 °F - closed cup

Flash Point(C)

-12.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, spectroscopic characterization, and reactivity of water-tolerant Eu3+-based precatalysts
Averill DJ and Allen MJ
Inorganic Chemistry, 53(12), 6257-6263 (2014)
Synthesis of (+)-and (−)-homononactic acid from (S)-1, 2-epoxybutane. Total synthesis of tetranactin by ′reverse coupe du roi′
Schmidt U and Werner J
Journal of the Chemical Society. Chemical Communications, 996-998 (1986)
A flexible synthesis of the phytoprostanes B1 type I and II
El Fangour S, et al.
The Journal of Organic Chemistry, 70(3), 989-997 (2005)
A Ohnishi et al.
Environmental research, 60(2), 242-247 (1993-02-01)
Axonal neuropathy occurs due to occupational ethylene oxide (EtO) exposure. The experimental model of human EtO neuropathy was established. In addition, the neurotoxic effects of propylene oxide (PpO) and butylene oxide (BtO) were demonstrated in rats. Although no human neuropathy
D Henschler et al.
Journal of cancer research and clinical oncology, 107(3), 149-156 (1984-01-01)
Previous analytical studies of industrial samples of trichloroethylene (TRI) have revealed the presence of mutagenic and carcinogenic epoxides which, it was proposed, might be responsible for the carcinogenicity of such samples, as demonstrated with mice in other laboratories. To test

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service