Skip to Content
Merck
All Photos(1)

Key Documents

532355

Sigma-Aldrich

(R)-(+)-1,2-Epoxybutane

98%

Synonym(s):

(2R)-Ethyloxirane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O
CAS Number:
Molecular Weight:
72.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D 10°, neat

refractive index

n20/D 1.386 (lit.)

bp

63 °C (lit.)

density

0.837 g/mL at 25 °C (lit.)

SMILES string

CC[C@@H]1CO1

InChI

1S/C4H8O/c1-2-4-3-5-4/h4H,2-3H2,1H3/t4-/m1/s1

InChI key

RBACIKXCRWGCBB-SCSAIBSYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

10.0 °F - closed cup

Flash Point(C)

-12.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Yuan Sun et al.
Carbohydrate research, 344(15), 1999-2004 (2009-08-26)
A new soluble cyclodextrin derivative 6-O-(2-hydroxybutyl)-beta-cyclodextrin (6-HB-beta-CD) was prepared. Its molecular binding and recognition ability were investigated with the comparison of beta-cyclodextrin (beta-CD), 2-O-(2-hydroxypropyl)-beta-cyclodextrin (2-HP-beta-CD), 6-O-(2-hydroxypropyl)-beta-cyclodextrin (6-HP-beta-CD), and 2-O-(2-hydroxybutyl)-beta-cyclodextrin (2-HB-beta-CD). The relationship between the complex stability constants and the possible
M Katz et al.
Journal of environmental pathology and toxicology, 3(5-6), 171-187 (1980-06-01)
Nitrosopiperidine, sodium nitrite and 1,2 epoxybutane were tested in the Ames agar incorporation assay in an attempt to establish exact criteria for detecting the activity of these weak mutagens. As regards minimum concentrations it was determined that at 500 microgram
1,2-Epoxybutane.
IARC monographs on the evaluation of carcinogenic risks to humans, 47, 217-228 (1989-01-01)
A M Rossi et al.
Teratogenesis, carcinogenesis, and mutagenesis, 3(1), 75-87 (1983-01-01)
In vivo and in vitro methodologies that have employed the yeast Schizosaccharomyces pombe as genetic indicator have been utilized to investigate the mutagenicity of two trichloroethylene (TCE) samples of pure and technical grade. Mutagenicity assays were also performed on two
Zhengyuan Zhou et al.
International journal of pharmaceutics, 354(1-2), 82-87 (2007-12-08)
Ethylene oxide and 1,2-butylene oxide were sequentially polymerised to form the diblock copolymer E13B10 (E=oxyethylene, B=oxybutylene, subscripts denote number-average block lengths in repeat units). Dynamic and static light scattering over the temperature range 10-30 degrees C demonstrated a transition from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service