Skip to Content
Merck
All Photos(1)

Key Documents

528056

Sigma-Aldrich

Thioglycolic acid

≥99%

Synonym(s):

Mercaptoacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COOH
CAS Number:
Molecular Weight:
92.12
Beilstein:
506166
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.2 (vs air)

Quality Level

vapor pressure

0.4 mmHg ( 25 °C)

Assay

≥99%

autoignition temp.

662 °F

refractive index

n20/D 1.505 (lit.)

bp

96 °C/5 mmHg (lit.)

mp

−16 °C (lit.)

density

1.326 g/mL at 20 °C (lit.)

functional group

carboxylic acid
thiol

storage temp.

2-8°C

SMILES string

OC(=O)CS

InChI

1S/C2H4O2S/c3-2(4)1-5/h5H,1H2,(H,3,4)

InChI key

CWERGRDVMFNCDR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Thioglycolic acid (TGA) can be used as:
  • A sulfur source and stabilizing agent to synthesize nanocrystalline metal sulfides via hydrothermal reaction. TGA also controls the morphology and particle sizes of nanocrystalline metal sulfides during the process.
  • An effective agglomeration suppressant for the agglomeration of sulfurized mineral pyrite.
  • A reactant to prepare 2,3-disubstituted-thiazolidin-4-one derivatives by one-pot cyclo-condensation reaction with various aldehydes and amines using supported protic acid catalyst.
  • A capping or stabilizing agent to synthesize CdTe quantum dots with controllable photoluminescence wavelengths.

Legal Information

Product of Arkema

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

266.0 °F - closed cup

Flash Point(C)

130 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of oxidation and thioglycolic acid on separation of coal and pyrite by selective oil agglomeration
DRZYMALA J and Wheelock TD
International Journal of Coal Preparation and Utilization, 14(1-2), 1-11 (1994)
Pyrite suppression in oil agglomeration of coal
Drzymala J, et al.
Proceedings, 1005-1008 (1991)
Controllable synthesis and cell-imaging studies on CdTe quantum dots together capped by glutathione and thioglycolic acid
Tian J, et al.
Journal of Colloid and Interface Science, 336(2), 504-509 (2009)
Silica chloride (SiO2-Cl) catalyzed one pot synthesis of 2, 3-disubstituted-thiazolidin-4-one
Gokhale KM and Telvekar VN
Synthetic Communications, 50(9), 1396-1403 (2020)
Controllable synthesis of nanocrystalline CdS with different morphologies by hydrothermal process in the presence of thioglycolic acid.
Salavati-Niasari M, et al.
Chemical Engineering Journal, 145(2), 346-350 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service