Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thioglycolic acid
Synonym(s):
Mercaptoacetic acid, Thioglycolic acid
Linear Formula:
HSCH
2
COOH
CAS Number:
68-11-1
Molecular Weight:
92.12
EC Number:
200-677-4
Products
Product Category
building blocks (1)
organic acids (1)
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
528056
Thioglycolic acid,
≥99%
Expand
View Pricing
1.00700
Thioglycolic acid,
GR for analysis Reag. Ph Eur
Expand
View Pricing
8.22336
Thioglycolic acid,
for synthesis
Expand
View Pricing
T3758
Thioglycolic acid,
98%
Expand
View Pricing