Skip to Content
Merck
All Photos(2)

Key Documents

513210

Sigma-Aldrich

(S)-(−)-2-Methyl-2-propanesulfinamide

97%

Synonym(s):

(S)-(-)-tert-Butanesulfinamide, (S)-(-)-tert-Butyl sulfinamide, (S)-2-Methyl-2-propanesulfinamide, (S)-tert-Butanesulfinamide, (S)-tert-Butylsulfinamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CS(O)NH2
CAS Number:
Molecular Weight:
121.20
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D −4.5°, c = 1 in chloroform

mp

97-101 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)S(N)=O

InChI

1S/C4H11NOS/c1-4(2,3)7(5)6/h5H2,1-3H3/t7-/m0/s1

InChI key

CESUXLKAADQNTB-ZETCQYMHSA-N

Application

(S)-(-)-2-Methyl-2-propanesulfinamide may be used to develop benzofuran-based farnesyltransferase inhibitors as anti-cancer agents. It may also be used to prepare (20E)-N-[t-butyl-(S)-sulfinyl]-3β-(t-butyldimethylsilyloxy)-pregn-5-en-20-imine, an intermediate for the preparation of androgen receptor antagonists.
Can be readily transformed into P,N-sulfinyl imine ligands through condensation with aldehydes and ketones, which can undergo iridium-catalyzed asymmetric hydrogenation of olefins.
Chiral auxiliary used in an asymmetric preparation of trifluoroethylamines by conversion of trifluoroacetaldehyde to a chiral imine followed by treatment with an aryl lithium and acidic methanolysis.
Useful reagent for synthesizing chiral amines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

20-Aminosteroids as a novel class of selective and complete androgen receptor antagonists and inhibitors of prostate cancer cell growth.
Fousteris MA, et al.
Bioorganic & Medicinal Chemistry Letters, 18(19), 6960-6969 (2010)
Synthesis and structure-activity relationships of novel benzofuran farnesyltransferase inhibitors.
Asoh K, et al.
Bioorganic & Medicinal Chemistry Letters, 19(6), 1753-1757 (2009)
Vouy Linh Truong et al.
Organic letters, 9(4), 683-685 (2007-01-30)
Condensation of N-tert-butanesulfinamide (S)-1 with trifluoroacetaldehyde hydrate 2a afforded 2-methyl-N-(2,2,2-trifluoroethylidene)propane-2-sulfinamide 3. Without isolation and purification, imine 3 was added to various aryllithium reagents to give highly diastereomerically enriched adducts 5a-g. Acidic methanolysis of 5a-g provided the desired 1-aryl-2,2,2-trifluoroethylamine hydrochloride compounds
Jeffrey P McMahon et al.
Organic letters, 6(10), 1645-1647 (2004-05-07)
Addition of alkyl or aryl Grignard reagents to N-sulfinyl imines derived from 3- and 4-substituted cyclohexanones proceeds with good yields and with excellent diasteroselectivity. The selectivity of the reaction is controlled by the ring substituent rather than the sulfinyl group
Journal of the American Chemical Society, 119, 9913-9914 (1997)

Articles

Ellman's sulfinamide is available in both enantiomeric and racemic forms for your research. This versatile and useful auxiliary has found extensive use both in academics and industry.

Ellman's sulfinamide is available in both enantiomeric and racemic forms for your research. This versatile and useful auxiliary has found extensive use both in academics and industry.

Ellman's sulfinamide is available in both enantiomeric and racemic forms for your research. This versatile and useful auxiliary has found extensive use both in academics and industry.

Ellman's sulfinamide is available in both enantiomeric and racemic forms for your research. This versatile and useful auxiliary has found extensive use both in academics and industry.

Related Content

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Ellman group developed electron-rich phosphine ligands for C-H functionalization and tert-Butanesulfinamide for asymmetric amine synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service