Skip to Content
Merck
All Photos(1)

Key Documents

491136

Sigma-Aldrich

1,4-Diaminobutane-2,2,3,3-d4 dihydrochloride

98 atom % D

Synonym(s):

1,4-Butanediamine-2,2,3,3-d4 dihydrochloride, Putrescine-2,2,3,3-d4 dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CD2CD2CH2NH2·2HCl
CAS Number:
Molecular Weight:
165.10
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

form

solid

mp

280 °C (dec.) (lit.)

mass shift

M+4

SMILES string

Cl.Cl.[2H]C([2H])(CN)C([2H])([2H])CN

InChI

1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H/i1D2,2D2;;

InChI key

XXWCODXIQWIHQN-RIZDZYNXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dana M Hardbower et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(5), E751-E760 (2017-01-18)
Macrophage activation is a critical step in host responses during bacterial infections. Ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine metabolism, has been well studied in epithelial cells and is known to have essential roles in many different cellular functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service