Skip to Content
Merck
All Photos(2)

Documents

D23602

Sigma-Aldrich

1,3-Diaminopropane

≥99%

Synonym(s):

1,3-Propanediamine, DAP, TMEDA, Trimethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3NH2
CAS Number:
Molecular Weight:
74.12
Beilstein:
605277
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<8 mmHg ( 20 °C)

Quality Level

Assay

≥99%

form

liquid

autoignition temp.

662 °F

expl. lim.

15.2 %

refractive index

n20/D 1.458 (lit.)

bp

140 °C (lit.)

mp

−12 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

SMILES string

NCCCN

InChI

1S/C3H10N2/c4-2-1-3-5/h1-5H2

InChI key

XFNJVJPLKCPIBV-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

1,3-Diaminopropane is used as a monomer for the synthesis of polyamides and aliphatic polyimides. It is also utilized for the modification of carbon nanotubes and serves as a cross-linking agent in the preparation of hydrogels.

Application

  • Synthesis and Characterization of Bulky 1,3-Diamidopropane Complexes of Group 2 Metals (Be ‒ Sr).: Demonstrates the utility of 1,3-diaminopropane in the synthesis of metal complexes, enhancing the diversity and functionality of inorganic compounds, crucial for high purity chemical reagent applications (Jones et al., 2024).
  • Rational engineering of homospermidine synthase for enhanced catalytic efficiency toward spermidine synthesis.: Explores the use of 1,3-diaminopropane as a precursor in enzymatic synthesis, optimizing biotechnological processes for polyamine production, pertinent to drug delivery system research (Liu et al., 2024).
  • Hydroxyl-terminated dendrimers with sulfonimide linkers as binders for metals of industrial significance.: Investigates 1,3-diaminopropane′s efficacy in the synthesis of dendrimers, crucial for industrial applications in binding metals, thereby impacting industrial chemical manufacturing R&D (Abu Sbeih et al., 2024).

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Resp. Sens. 1B - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

122.0 °F

Flash Point(C)

50 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinfeng Ding et al.
Frontiers in microbiology, 8, 1842-1842 (2017-10-12)
Diamine biosynthesis: research progress and application prospects
Wang et al.
Applied and Environmental Microbiology, 86, e01972-e01920 (2020)
Low-and high-resolution nuclear magnetic resonance (NMR) characterisation of hyaluronan-based native and sulfated hydrogels
Barbucci et al.
Carbohydrate Research, 341 (2006)
Xiaoyue He et al.
Nature communications, 10(1), 2847-2847 (2019-06-30)
The control of the density and type of line defects on two-dimensional (2D) materials enable the development of new methods to tailor their physical and chemical properties. In particular, mirror twin boundaries (MTBs) on transition metal dichacogenides have attracted much
Eleftheria Soulioti et al.
Medicina (Kaunas, Lithuania), 55(7) (2019-07-26)
Background and objectives: Brachial plexus block is commonly used in shoulder surgery, as it provides satisfactory surgical conditions and adequate postoperative pain control. However, there are contradictory reports regarding the addition of tramadol to the injected regional anesthetic solution. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service