Skip to Content
Merck
All Photos(1)

Key Documents

490490

Sigma-Aldrich

Perylene-d12

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20D12
CAS Number:
Molecular Weight:
264.38
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

form

solid

mp

277-279 °C (lit.)

application(s)

electronics

mass shift

M+12

SMILES string

[2H]c1c([2H])c2c([2H])c([2H])c([2H])c3c4c([2H])c([2H])c([2H])c5c([2H])c([2H])c([2H])c(c(c1[2H])c23)c45

InChI

1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D

InChI key

CSHWQDPOILHKBI-AQZSQYOVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng-Xia Chen et al.
Chemosphere, 131, 192-200 (2015-04-12)
Fine roots of woody plants are a heterogeneous system differing markedly in structure and function. Nevertheless, knowledge about the plant uptake of organic pollutants via fine roots is scarce to date. In the present study, plant uptake, translocation, and return
Chen-Chou Wu et al.
Environmental science & technology, 49(17), 10607-10615 (2015-08-11)
Barbecuing or charcoal-grilling has become part of popular outdoor recreational activities nowadays; however, potential human health hazards through outdoor exposure to barbecue fumes have yet to be adequately quantified. To fill this knowledge gap, atmospheric size-fractioned particle and gaseous samples
Fereshteh Shahhoseini et al.
Talanta, 207, 120320-120320 (2019-10-09)
The need for high throughput, reliable analytical methods for environmental analysis has driven our development of novel sampling technologies that can be used in microextraction devices and integrated into chromatographic systems for fast analysis. Here we report a new method
Ali Azizi et al.
Journal of chromatography. A, 1610, 460534-460534 (2019-09-22)
Magnetic molecularly imprinted polymers (MMIPs) combine nanotechnology and molecular imprinting technology to offer selective and tunable enrichment for water analysis. In this paper, a selective sorbent was prepared by surface polymerization onto magnetic Fe3O4@SiO2 nanoparticles through reversible addition fragmentation chain
Haihua Jiao et al.
International journal of environmental research and public health, 12(6), 5775-5791 (2015-05-29)
The levels of 16 polycyclic aromatic hydrocarbons (PAHs) were investigated in 27 upper layer (0-25 cm) soil samples collected from the Dagang Oilfield (China) in April 2013 to estimate their distribution, possible sources, and potential risks posed. The total concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service