Skip to Content
Merck
All Photos(1)

Key Documents

451819

Sigma-Aldrich

Acenaphthene-d10

99 atom % D

Synonym(s):

1,8-·Ethylenenaphthalene-D10, Deuterated acenaphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D10
CAS Number:
Molecular Weight:
164.27
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

solid

bp

277-279 °C (lit.)

mp

95-97 °C (lit.)

mass shift

M+10

SMILES string

[2H]c1c([2H])c2c3c(c([2H])c([2H])c([2H])c3c1[2H])C([2H])([2H])C2([2H])[2H]

InChI

1S/C12H10/c1-3-9-4-2-6-11-8-7-10(5-1)12(9)11/h1-6H,7-8H2/i1D,2D,3D,4D,5D,6D,7D2,8D2

InChI key

CWRYPZZKDGJXCA-WHUVPORUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

May be used as an analytical standard

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Chou Wu et al.
Environmental science & technology, 49(17), 10607-10615 (2015-08-11)
Barbecuing or charcoal-grilling has become part of popular outdoor recreational activities nowadays; however, potential human health hazards through outdoor exposure to barbecue fumes have yet to be adequately quantified. To fill this knowledge gap, atmospheric size-fractioned particle and gaseous samples
Noriatsu Ozaki et al.
Water research, 80, 337-345 (2015-06-01)
The loading and removal of polycyclic aromatic hydrocarbons (PAHs) were measured and estimated in a wastewater treatment plant in a separated sewer system in a suburban area of Japan. The influent 16 PAHs concentration was 219 ± 210 ng L(-1)
Nien-Hsin Kao et al.
Environmental science and pollution research international, 22(10), 7529-7546 (2015-02-26)
The sources of the spill and the contaminated soils of an aged oil spill contaminated site with unknown mixed pollutants were investigated by using a set of developed forensic chemical procedures which include analysis of oil products, site investigation, gas
Tirumuru V Reddy et al.
Environmental toxicology and chemistry, 34(12), 2796-2802 (2015-06-20)
Endocrine-disrupting chemicals (EDCs) such as 17α-ethynylestradiol, 17β-estradiol, estrone, and para-nonylphenol have been measured in wastewater-treatment plant effluents, surface waters, sediments, and sludge and have been shown to induce liver-specific vitellogenin (vtg) messenger RNA in male fathead minnows (Pimephales promelas). The
Nien-Hsin Kao et al.
Marine pollution bulletin, 97(1-2), 319-332 (2015-06-08)
Three fishing harbors were investigated to study the polycyclic aromatic hydrocarbons in the sediments and trace possible anthropogenic sources by identification of cyclic terpenoid biomarkers. Seventeen terpanes, 10 steranes and 10 bicyclic sesquiterpanes in the marine diesel and the three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service