Skip to Content
Merck
All Photos(1)

Key Documents

488100

Sigma-Aldrich

Aniline-15N

98 atom % 15N

Synonym(s):

Benzenamine-15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H515NH2
CAS Number:
Molecular Weight:
94.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 15N

Assay

99% (CP)

refractive index

n20/D 1.586 (lit.)

bp

184 °C (lit.)

mp

−6 °C (lit.)

density

1.033 g/mL at 25 °C

mass shift

M+1

storage temp.

2-8°C

SMILES string

[15NH2]c1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2/i7+1

InChI key

PAYRUJLWNCNPSJ-CDYZYAPPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Resp. Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conghui Tang et al.
Journal of the American Chemical Society, 134(46), 18924-18927 (2012-11-08)
A novel and efficient copper-catalyzed azidation reaction of anilines via C-H activation has been developed. This method, in which the primary amine acts as a directing group by coordinating to the metal center, provides ortho azidation products regioselectively under mild
Sung Jin Bae et al.
European journal of medicinal chemistry, 57, 383-390 (2012-11-15)
We attempted to design and synthesize (E)-N-substituted benzylidene-hydroxy or methoxy-aniline derivatives and to evaluate their inhibitory effect on tyrosinase activity and anti-melanogenesis activity in murine B16F10 melanoma cells. Derivatives with a 4-methoxy- or 4-hydroxy-anilino group exerted more potent inhibition against
Huihui Yang et al.
Electrophoresis, 34(4), 510-517 (2012-11-23)
A novel (3-sulfopropyl methacrylate potassium)-silica hybrid monolithic column for CEC has been prepared by a simple one-pot approach based on efficient thiol-ene click chemistry. In this process, the polycondensation of hydrolyzed alkoxysilanes and in situ click reaction of vinyl groups
Yu Qian et al.
Chemical communications (Cambridge, England), 49(26), 2700-2702 (2013-02-26)
An enantioselective four-component reaction of a diazoketone, water, an aniline and ethyl glyoxylate in the presence of catalytic Rh2(OAc)4 and a chiral Brønsted acid was developed to efficiently produce β-hydroxy-α-amino acid derivatives in good yields with high diastereoselectivity and enantioselectivity.
Rongjun He et al.
Chemical communications (Cambridge, England), 49(20), 2064-2066 (2013-02-06)
Mycobacterium protein tyrosine phosphatase B (mPTPB) is essential for the survival and persistence of Mycobacterium in the host. Thus small molecule inhibitors of mPTPB are potential anti-TB agents. We developed an efficient organocatalytic multicomponent reaction (MCR) between pyrrole, formaldehyde and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service