Merck
  • A highly enantioselective four-component reaction for the efficient construction of chiral β-hydroxy-α-amino acid derivatives.
A highly enantioselective four-component reaction for the efficient construction of chiral β-hydroxy-α-amino acid derivatives.
Chemical communications (Cambridge, England) (2013-02-26)
Yu Qian, Changcheng Jing, Shunying Liu, Wenhao Hu
ABSTRACT
An enantioselective four-component reaction of a diazoketone, water, an aniline and ethyl glyoxylate in the presence of catalytic Rh2(OAc)4 and a chiral Brønsted acid was developed to efficiently produce β-hydroxy-α-amino acid derivatives in good yields with high diastereoselectivity and enantioselectivity.
Sigma-Aldrich
Aniline, SAJ first grade, ≥99.0%
Sigma-Aldrich
Aniline, JIS special grade, ≥99.0%
Sigma-Aldrich
Aniline-4-13C, 99 atom % 13C
Supelco
Aniline, analytical standard
Sigma-Aldrich
Aniline, ReagentPlus®, 99%
Sigma-Aldrich
Aniline-15N, 98 atom % 15N
Sigma-Aldrich
Aniline, ACS reagent, ≥99.5%
Sigma-Aldrich
Aniline-1-13C, 99 atom % 13C
Sigma-Aldrich
Aniline hydrochloride, ≥99%
Supelco
Aniline solution, certified reference material, 5000 μg/mL in methanol