Skip to Content
Merck
All Photos(1)

Key Documents

480959

Sigma-Aldrich

Glycolamide

98%

Synonym(s):

2-Hydroxyacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CONH2
CAS Number:
Molecular Weight:
75.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

118-120 °C (lit.)

functional group

amide
hydroxyl

SMILES string

NC(=O)CO

InChI

1S/C2H5NO2/c3-2(5)1-4/h4H,1H2,(H2,3,5)

InChI key

TZGPACAKMCUCKX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycolamide, an isomer of glycine, can be synthesized by the ammonolysis of ethyl glycolate. The conformational analysis of glycolamide has been reported. Its diffusion coefficient in water and partial molal isothermal compressibility has been determined. Its thermodynamic properties have been investigated. Its osmotic and activity coefficients have been obtained.

Application

Glycolamide may be used in the preparation of phosphoglycolamide.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Sean D Reilly et al.
Chemical communications (Cambridge, England), 48(78), 9732-9734 (2012-08-24)
Complexation of Pu(IV) with TMDGA, TEDGA, and TODGA diglycolamide ligands was followed by vis-NIR spectroscopy. A crystal structure determination reveals that TMDGA forms a 1 : 3 homoleptic Pu(IV) complex with the nitrate anions forced into the outer coordination sphere.
Parmeshwari K Halen et al.
Mini reviews in medicinal chemistry, 9(1), 124-139 (2009-01-20)
Non-steroidal anti-inflammatory drugs (NSAIDs), commonly used for the treatment of chronic inflammatory diseases suffer from several undesired side effects, the most important being gastrointestinal (GI) irritation and ulceration. The prodrug designing is one of the several strategies used to overcome
Partial molal isothermal compressibility of glycine, DL-α-alanine, glycylglycine, glycolamide, and lactamide.
Yayanos AA.
The Journal of Physical Chemistry, 97(49), 13027-13028 (1993)
Fereidoon Daryaee et al.
European journal of medicinal chemistry, 44(1), 289-295 (2008-04-26)
On the basis of the structural similarity of N-substituted glycolamides with N-glycolyl muramic acid residues of the cell wall of Mycobacterium tuberculosis, a series of these compounds were designed and synthesized by the reaction of glycolic acid acetonide 1 (2,2-dimethyl-5-oxo-1,3-dioxolane)
A K Bansal et al.
Drug development and industrial pharmacy, 27(1), 63-70 (2001-03-15)
Glycolamide esters of ibuprofen (I), namely, unsubstituted (II), N,N dimethyl (III), and N,N diethyl (IV), were synthesized and studied for different physicochemical, pharmacological, and toxicological properties. They were comparable with I in respect of anti-inflammatory and analgesic activity but did

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service